Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11241412-11241673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL8
Entrez Gene ID
55908
Gene Name
angiopoietin like 8
Gene Aliases
C19orf80, PRO1185, PVPA599, RIFL, TD26
UniGene
Hs.534467
Gene Chromosome Location
Chr.19: 11239619-11241943 on Build GRCh38
Transcripts
NM_018687.6 NM_020812.3 XM_005260000.2 XM_005260001.2 XM_011528150.1 XM_011528151.1 XM_011528152.1

Gene Symbol
DOCK6
Entrez Gene ID
57572
Gene Name
dedicator of cytokinesis 6
Gene Aliases
AOS2, ZIR1
UniGene
Hs.591002
Gene Chromosome Location
Chr.19: 11199293-11262502 on Build GRCh38
Transcripts
NM_018687.6 NM_020812.3 XM_005260000.2 XM_005260001.2 XM_011528150.1 XM_011528151.1 XM_011528152.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4445453 c.569+3G>A p.? Chr.19: 11241557-11241557 on Build GRCh38
5520932 c.465C>T p.H155H Chr.19: 11241450-11241450 on Build GRCh38
5548915 c.535G>A p.A179T Chr.19: 11241520-11241520 on Build GRCh38
4938453 c.496C>T p.L166F Chr.19: 11241481-11241481 on Build GRCh38
6397046 c.496C>A p.L166I Chr.19: 11241481-11241481 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768124706 Chr.19: 11241500-11241500 on Build GRCh38
rs577165851 Chr.19: 11241534-11241534 on Build GRCh38
rs541695168 Chr.19: 11241636-11241636 on Build GRCh38
rs145750006 Chr.19: 11241647-11241647 on Build GRCh38
rs760883807 Chr.19: 11241567-11241567 on Build GRCh38
rs896572226 Chr.19: 11241492-11241492 on Build GRCh38
rs375004104 Chr.19: 11241493-11241493 on Build GRCh38
rs192460764 Chr.19: 11241499-11241499 on Build GRCh38
rs772769447 Chr.19: 11241554-11241554 on Build GRCh38
rs952350168 Chr.19: 11241607-11241607 on Build GRCh38
rs778603926 Chr.19: 11241451-11241451 on Build GRCh38
rs781506138 Chr.19: 11241508-11241508 on Build GRCh38
rs957168765 Chr.19: 11241579-11241579 on Build GRCh38
rs371251856 Chr.19: 11241480-11241480 on Build GRCh38
rs559730290 Chr.19: 11241597-11241597 on Build GRCh38
rs539791974 Chr.19: 11241501-11241501 on Build GRCh38
rs759926768 Chr.19: 11241539-11241539 on Build GRCh38
rs753983886 Chr.19: 11241438-11241438 on Build GRCh38
rs535690161 Chr.19: 11241474-11241474 on Build GRCh38
rs750243179 Chr.19: 11241637-11241637 on Build GRCh38
rs557880890 Chr.19: 11241528-11241528 on Build GRCh38
rs757450296 Chr.19: 11241439-11241439 on Build GRCh38
rs1007339147 Chr.19: 11241509-11241509 on Build GRCh38
rs377337313 Chr.19: 11241600-11241600 on Build GRCh38
rs776106774 Chr.19: 11241559-11241559 on Build GRCh38
rs541334378 Chr.19: 11241543-11241543 on Build GRCh38
rs1019024305 Chr.19: 11241547-11241547 on Build GRCh38
rs985471363 Chr.19: 11241609-11241609 on Build GRCh38
rs940570679 Chr.19: 11241459-11241459 on Build GRCh38
rs746610288 Chr.19: 11241654-11241654 on Build GRCh38
rs764289104 Chr.19: 11241590-11241590 on Build GRCh38
rs761944598 Chr.19: 11241603-11241603 on Build GRCh38
rs774587241 Chr.19: 11241538-11241538 on Build GRCh38
rs146610047 Chr.19: 11241648-11241648 on Build GRCh38
rs568320463 Chr.19: 11241443-11241443 on Build GRCh38
rs747781883 Chr.19: 11241532-11241532 on Build GRCh38
rs1020159418 Chr.19: 11241601-11241601 on Build GRCh38

View Full Product Details