Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8836190-8836415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A2ML1
Entrez Gene ID
144568
Gene Name
alpha-2-macroglobulin like 1
Gene Aliases
CPAMD9
UniGene
Hs.620532
Gene Chromosome Location
Chr.12: 8822472-8887202 on Build GRCh38
Transcripts
NM_144670.5 XM_011520566.2 XM_011520567.2 XM_017018868.1 XM_017018869.1 XM_017018870.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5418489 c.644-10C>G p.? Chr.12: 8836245-8836245 on Build GRCh38
7227998 c.670G>T p.V224L Chr.12: 8836281-8836281 on Build GRCh38
7353360 c.682G>T p.E228* Chr.12: 8836293-8836293 on Build GRCh38
7239866 c.708C>G p.F236L Chr.12: 8836319-8836319 on Build GRCh38
4556174 c.681G>A p.K227K Chr.12: 8836292-8836292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373095814 Chr.12: 8836352-8836352 on Build GRCh38
rs748686815 Chr.12: 8836292-8836292 on Build GRCh38
rs369163411 Chr.12: 8836239-8836239 on Build GRCh38
rs776997039 Chr.12: 8836374-8836374 on Build GRCh38
rs992780546 Chr.12: 8836312-8836312 on Build GRCh38
rs374554974 Chr.12: 8836347-8836347 on Build GRCh38
rs761197510 Chr.12: 8836262-8836262 on Build GRCh38
rs773797927 Chr.12: 8836261-8836261 on Build GRCh38
rs751739174 Chr.12: 8836391-8836391 on Build GRCh38
rs766575041 Chr.12: 8836384-8836384 on Build GRCh38
rs748034976 Chr.12: 8836325-8836325 on Build GRCh38
rs755006090 Chr.12: 8836320-8836320 on Build GRCh38
rs964036663 Chr.12: 8836394-8836394 on Build GRCh38
rs778770820 Chr.12: 8836375-8836376 on Build GRCh38
rs750760910 Chr.12: 8836228-8836228 on Build GRCh38
rs765561612 Chr.12: 8836290-8836290 on Build GRCh38
rs1055961822 Chr.12: 8836308-8836308 on Build GRCh38
rs767846073 Chr.12: 8836365-8836365 on Build GRCh38
rs377719401 Chr.12: 8836364-8836364 on Build GRCh38
rs192636548 Chr.12: 8836395-8836395 on Build GRCh38
rs763298807 Chr.12: 8836383-8836383 on Build GRCh38
rs202208600 Chr.12: 8836253-8836253 on Build GRCh38
rs199682918 Chr.12: 8836304-8836304 on Build GRCh38
rs777100151 Chr.12: 8836282-8836282 on Build GRCh38
rs370100637 Chr.12: 8836368-8836368 on Build GRCh38
rs772728105 Chr.12: 8836252-8836252 on Build GRCh38
rs969579536 Chr.12: 8836330-8836330 on Build GRCh38
rs200465216 Chr.12: 8836323-8836323 on Build GRCh38
rs769537819 Chr.12: 8836250-8836250 on Build GRCh38
rs762067269 Chr.12: 8836283-8836283 on Build GRCh38
rs201550299 Chr.12: 8836269-8836269 on Build GRCh38
rs773576690 Chr.12: 8836380-8836380 on Build GRCh38
rs765531790 Chr.12: 8836379-8836379 on Build GRCh38
rs777559324 Chr.12: 8836335-8836335 on Build GRCh38
rs762311589 Chr.12: 8836376-8836376 on Build GRCh38
rs755860036 Chr.12: 8836332-8836332 on Build GRCh38
rs202211381 Chr.12: 8836303-8836303 on Build GRCh38
rs756810420 Chr.12: 8836265-8836265 on Build GRCh38
rs946045239 Chr.12: 8836232-8836232 on Build GRCh38
rs774943263 Chr.12: 8836280-8836280 on Build GRCh38
rs750612606 Chr.12: 8836291-8836291 on Build GRCh38
rs770435207 Chr.12: 8836254-8836254 on Build GRCh38
rs144841693 Chr.12: 8836216-8836216 on Build GRCh38

View Full Product Details