Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42188622-42188867 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXJ3
Entrez Gene ID
22887
Gene Name
forkhead box J3
Gene Aliases
-
UniGene
Hs.26023
Gene Chromosome Location
Chr.1: 42176539-42335877 on Build GRCh38
Transcripts
NM_001198850.1 NM_001198851.1 NM_001198852.1 NM_014947.4 XM_005270632.4 XM_006710458.3 XM_006710459.3 XM_011541026.2 XM_017000693.1 XM_017000694.1 XM_017000695.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7368651 c.1525T>C p.C509R Chr.1: 42188857-42188857 on Build GRCh38
2169747 c.1534C>T p.L512F Chr.1: 42188848-42188848 on Build GRCh38
4528188 c.1519G>A p.D507N Chr.1: 42188863-42188863 on Build GRCh38
6126728 c.1542G>T p.Q514H Chr.1: 42188840-42188840 on Build GRCh38
6666296 c.1517C>T p.A506V Chr.1: 42188865-42188865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761180457 Chr.1: 42188730-42188730 on Build GRCh38
rs757960670 Chr.1: 42188807-42188807 on Build GRCh38
rs892849842 Chr.1: 42188692-42188692 on Build GRCh38
rs901322085 Chr.1: 42188818-42188818 on Build GRCh38
rs935665192 Chr.1: 42188745-42188745 on Build GRCh38
rs11799854 Chr.1: 42188688-42188688 on Build GRCh38
rs780195657 Chr.1: 42188797-42188797 on Build GRCh38
rs762457256 Chr.1: 42188717-42188717 on Build GRCh38
rs758657783 Chr.1: 42188791-42188791 on Build GRCh38
rs573579304 Chr.1: 42188719-42188719 on Build GRCh38
rs780942907 Chr.1: 42188817-42188817 on Build GRCh38
rs942857220 Chr.1: 42188754-42188754 on Build GRCh38
rs754807643 Chr.1: 42188815-42188815 on Build GRCh38
rs58227404 Chr.1: 42188837-42188837 on Build GRCh38
rs554500 Chr.1: 42188693-42188693 on Build GRCh38
rs372696955 Chr.1: 42188689-42188689 on Build GRCh38
rs138469226 Chr.1: 42188781-42188781 on Build GRCh38
rs765384065 Chr.1: 42188759-42188759 on Build GRCh38
rs1051312276 Chr.1: 42188660-42188660 on Build GRCh38
rs754440979 Chr.1: 42188744-42188744 on Build GRCh38
rs910257077 Chr.1: 42188752-42188752 on Build GRCh38
rs757819651 Chr.1: 42188751-42188751 on Build GRCh38
rs564684670 Chr.1: 42188802-42188802 on Build GRCh38
rs568102835 Chr.1: 42188718-42188718 on Build GRCh38
rs762699572 Chr.1: 42188702-42188702 on Build GRCh38
rs115993725 Chr.1: 42188681-42188681 on Build GRCh38
rs746161839 Chr.1: 42188722-42188722 on Build GRCh38
rs367714767 Chr.1: 42188734-42188734 on Build GRCh38
rs772546331 Chr.1: 42188696-42188696 on Build GRCh38
rs748104963 Chr.1: 42188831-42188831 on Build GRCh38
rs747223827 Chr.1: 42188814-42188814 on Build GRCh38

View Full Product Details