Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 206110510-206110237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVPR1B
Entrez Gene ID
553
Gene Name
arginine vasopressin receptor 1B
Gene Aliases
AVPR3
UniGene
Hs.1372
Gene Chromosome Location
Chr.1: 206109849-206117048 on Build GRCh38
Transcripts
NM_000707.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2215774 c.1090C>T p.R364C Chr.1: 206110374-206110374 on Build GRCh38
2215772 c.1069G>T p.G357W Chr.1: 206110395-206110395 on Build GRCh38
6608973 c.1041G>A p.P347P Chr.1: 206110423-206110423 on Build GRCh38
4985678 c.1091G>A p.R364H Chr.1: 206110373-206110373 on Build GRCh38
4923464 c.1030C>A p.H344N Chr.1: 206110434-206110434 on Build GRCh38
View More

View Full Product Details