Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 35078714-35078977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM177A1
Entrez Gene ID
283635
Gene Name
family with sequence similarity 177 member A1
Gene Aliases
C14orf24
UniGene
Hs.446357
Gene Chromosome Location
Chr.14: 35044907-35083383 on Build GRCh38
Transcripts
NM_001079519.1 NM_001289022.1 NM_173607.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5342446 c.352G>A p.G118R Chr.14: 35078941-35078941 on Build GRCh38
5647279 c.437C>T p.S146F Chr.14: 35078957-35078957 on Build GRCh38
4836278 c.368C>G p.S123C Chr.14: 35078957-35078957 on Build GRCh38
4836277 c.437C>G p.S146C Chr.14: 35078957-35078957 on Build GRCh38
7367160 c.426G>C p.E142D Chr.14: 35078946-35078946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781638103 Chr.14: 35078883-35078883 on Build GRCh38
rs770338620 Chr.14: 35078913-35078914 on Build GRCh38
rs533654975 Chr.14: 35078753-35078753 on Build GRCh38
rs762079334 Chr.14: 35078901-35078902 on Build GRCh38
rs980751474 Chr.14: 35078850-35078850 on Build GRCh38
rs938358461 Chr.14: 35078757-35078757 on Build GRCh38
rs1042904546 Chr.14: 35078831-35078831 on Build GRCh38
rs1008664814 Chr.14: 35078906-35078906 on Build GRCh38
rs747499880 Chr.14: 35078864-35078864 on Build GRCh38
rs773914706 Chr.14: 35078794-35078794 on Build GRCh38
rs770483938 Chr.14: 35078898-35078898 on Build GRCh38
rs956038192 Chr.14: 35078756-35078756 on Build GRCh38
rs748682373 Chr.14: 35078892-35078892 on Build GRCh38
rs770394464 Chr.14: 35078750-35078750 on Build GRCh38
rs200183172 Chr.14: 35078912-35078912 on Build GRCh38
rs368802731 Chr.14: 35078899-35078899 on Build GRCh38
rs917506221 Chr.14: 35078761-35078761 on Build GRCh38
rs748844205 Chr.14: 35078908-35078908 on Build GRCh38
rs148889643 Chr.14: 35078752-35078752 on Build GRCh38
rs770321512 Chr.14: 35078915-35078915 on Build GRCh38
rs774072355 Chr.14: 35078946-35078946 on Build GRCh38
rs947226905 Chr.14: 35078804-35078804 on Build GRCh38
rs769056164 Chr.14: 35078876-35078876 on Build GRCh38
rs28496323 Chr.14: 35078819-35078819 on Build GRCh38
rs751683724 Chr.14: 35078878-35078878 on Build GRCh38
rs370372725 Chr.14: 35078880-35078880 on Build GRCh38
rs948962899 Chr.14: 35078762-35078762 on Build GRCh38
rs992802592 Chr.14: 35078759-35078759 on Build GRCh38
rs773545030 Chr.14: 35078937-35078937 on Build GRCh38
rs748984227 Chr.14: 35078740-35078740 on Build GRCh38

View Full Product Details