Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50378509-50378772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKK
Entrez Gene ID
139189
Gene Name
diacylglycerol kinase kappa
Gene Aliases
-
UniGene
Hs.38132
Gene Chromosome Location
Chr.X: 50365407-50470850 on Build GRCh38
Transcripts
NM_001013742.3 XM_017029268.1

SNP Detail

SNP ID Chromosome Location
rs782755784 Chr.X: 50378697-50378697 on Build GRCh38
rs782545185 Chr.X: 50378738-50378738 on Build GRCh38
rs781913087 Chr.X: 50378723-50378723 on Build GRCh38
rs782265187 Chr.X: 50378530-50378530 on Build GRCh38
rs782283261 Chr.X: 50378566-50378566 on Build GRCh38
rs375861957 Chr.X: 50378581-50378581 on Build GRCh38
rs781933403 Chr.X: 50378625-50378625 on Build GRCh38
rs782512781 Chr.X: 50378532-50378532 on Build GRCh38
rs782048910 Chr.X: 50378635-50378635 on Build GRCh38
rs372923543 Chr.X: 50378545-50378545 on Build GRCh38
rs781924899 Chr.X: 50378578-50378578 on Build GRCh38
rs1038728010 Chr.X: 50378683-50378683 on Build GRCh38
rs781978930 Chr.X: 50378606-50378606 on Build GRCh38
rs782159939 Chr.X: 50378612-50378612 on Build GRCh38
rs369713639 Chr.X: 50378696-50378696 on Build GRCh38
rs781992907 Chr.X: 50378559-50378559 on Build GRCh38
rs782756939 Chr.X: 50378670-50378670 on Build GRCh38
rs782400492 Chr.X: 50378558-50378558 on Build GRCh38
rs782379283 Chr.X: 50378596-50378596 on Build GRCh38
rs781824281 Chr.X: 50378671-50378671 on Build GRCh38
rs782796172 Chr.X: 50378624-50378624 on Build GRCh38
rs782742321 Chr.X: 50378639-50378640 on Build GRCh38
rs182655837 Chr.X: 50378740-50378740 on Build GRCh38
rs782331960 Chr.X: 50378567-50378567 on Build GRCh38
rs934938048 Chr.X: 50378544-50378544 on Build GRCh38

View Full Product Details