Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38232770-38233043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDHD2
Entrez Gene ID
23259
Gene Name
DDHD domain containing 2
Gene Aliases
SAMWD1, SPG54, iPLA(1)gamma
UniGene
Hs.434966
Gene Chromosome Location
Chr.8: 38231343-38273442 on Build GRCh38
Transcripts
NM_001164232.1 NM_001164234.1 NM_015214.2 XM_005273454.2 XM_005273455.3 XM_005273456.3 XM_011544456.1 XM_017013252.1 XM_017013253.1 XM_017013254.1 XM_017013255.1 XM_017013256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6516099 c.5C>A p.S2* Chr.8: 38232999-38232999 on Build GRCh38
5746864 c.40T>C p.S14P Chr.8: 38233034-38233034 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779600306 Chr.8: 38232947-38232947 on Build GRCh38
rs905317841 Chr.8: 38232944-38232944 on Build GRCh38
rs771376379 Chr.8: 38233011-38233011 on Build GRCh38
rs773201028 Chr.8: 38232890-38232890 on Build GRCh38
rs958932942 Chr.8: 38232973-38232973 on Build GRCh38
rs770287920 Chr.8: 38232972-38232972 on Build GRCh38
rs192202566 Chr.8: 38232822-38232822 on Build GRCh38
rs746727123 Chr.8: 38232948-38232948 on Build GRCh38
rs7834485 Chr.8: 38232902-38232902 on Build GRCh38
rs7813547 Chr.8: 38232898-38232898 on Build GRCh38
rs1002973184 Chr.8: 38232932-38232933 on Build GRCh38
rs754417987 Chr.8: 38232950-38232950 on Build GRCh38
rs201334641 Chr.8: 38233013-38233013 on Build GRCh38
rs1015096121 Chr.8: 38232923-38232926 on Build GRCh38
rs573340745 Chr.8: 38232929-38232929 on Build GRCh38
rs768668773 Chr.8: 38232912-38232913 on Build GRCh38
rs772161301 Chr.8: 38232827-38232827 on Build GRCh38
rs986245942 Chr.8: 38232823-38232823 on Build GRCh38
rs7813415 Chr.8: 38232850-38232850 on Build GRCh38
rs977425695 Chr.8: 38233004-38233004 on Build GRCh38
rs199619753 Chr.8: 38232989-38232989 on Build GRCh38
rs919527123 Chr.8: 38232922-38232922 on Build GRCh38
rs780823905 Chr.8: 38232951-38232951 on Build GRCh38
rs972311370 Chr.8: 38232912-38232912 on Build GRCh38
rs776821835 Chr.8: 38232970-38232974 on Build GRCh38
rs7834509 Chr.8: 38232957-38232957 on Build GRCh38
rs762551995 Chr.8: 38232801-38232801 on Build GRCh38
rs748593776 Chr.8: 38232959-38232959 on Build GRCh38
rs760594462 Chr.8: 38233006-38233006 on Build GRCh38
rs888090435 Chr.8: 38232878-38232878 on Build GRCh38
rs7824699 Chr.8: 38232954-38232954 on Build GRCh38
rs997012528 Chr.8: 38232870-38232871 on Build GRCh38
rs941685463 Chr.8: 38232876-38232876 on Build GRCh38
rs1046564671 Chr.8: 38232936-38232936 on Build GRCh38
rs938146404 Chr.8: 38232992-38232992 on Build GRCh38
rs908919358 Chr.8: 38232843-38232843 on Build GRCh38
rs774404330 Chr.8: 38233012-38233012 on Build GRCh38
rs562818441 Chr.8: 38232977-38232977 on Build GRCh38
rs141540044 Chr.8: 38232871-38232873 on Build GRCh38
rs182341575 Chr.8: 38233015-38233015 on Build GRCh38
rs949609657 Chr.8: 38232933-38232933 on Build GRCh38

View Full Product Details