Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 27876411-27876658 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THEMIS2
Entrez Gene ID
9473
Gene Name
thymocyte selection associated family member 2
Gene Aliases
C1orf38, ICB-1
UniGene
Hs.10649
Gene Chromosome Location
Chr.1: 27872531-27886685 on Build GRCh38
Transcripts
NM_001039477.2 NM_001105556.2 NM_001286113.1 NM_001286115.1 NM_004848.3 XM_005246041.3 XM_006711050.1 XM_011542445.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6801211 c.137C>T p.T46M Chr.1: 27876630-27876630 on Build GRCh38
331190 c.102C>A p.I34I Chr.1: 27876595-27876595 on Build GRCh38
3671712 c.115G>T p.G39W Chr.1: 27876608-27876608 on Build GRCh38
6453688 c.138G>A p.T46T Chr.1: 27876631-27876631 on Build GRCh38
4850407 c.150C>T p.I50I Chr.1: 27876643-27876643 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749157499 Chr.1: 27876630-27876630 on Build GRCh38
rs957767444 Chr.1: 27876603-27876603 on Build GRCh38
rs199944155 Chr.1: 27876563-27876563 on Build GRCh38
rs759144236 Chr.1: 27876638-27876638 on Build GRCh38
rs747530055 Chr.1: 27876472-27876472 on Build GRCh38
rs913680334 Chr.1: 27876536-27876536 on Build GRCh38
rs764336829 Chr.1: 27876621-27876621 on Build GRCh38
rs952118391 Chr.1: 27876440-27876440 on Build GRCh38
rs191288059 Chr.1: 27876584-27876584 on Build GRCh38
rs377358709 Chr.1: 27876528-27876528 on Build GRCh38
rs112947097 Chr.1: 27876529-27876529 on Build GRCh38
rs758434367 Chr.1: 27876581-27876581 on Build GRCh38
rs975704576 Chr.1: 27876494-27876494 on Build GRCh38
rs371084168 Chr.1: 27876559-27876559 on Build GRCh38
rs535414354 Chr.1: 27876503-27876503 on Build GRCh38
rs774954578 Chr.1: 27876622-27876622 on Build GRCh38
rs756602762 Chr.1: 27876540-27876540 on Build GRCh38
rs199945403 Chr.1: 27876597-27876597 on Build GRCh38
rs753209297 Chr.1: 27876539-27876539 on Build GRCh38
rs139282726 Chr.1: 27876432-27876432 on Build GRCh38
rs1045895189 Chr.1: 27876546-27876546 on Build GRCh38
rs746101424 Chr.1: 27876577-27876577 on Build GRCh38
rs754241049 Chr.1: 27876561-27876561 on Build GRCh38
rs922472382 Chr.1: 27876452-27876452 on Build GRCh38
rs1018001876 Chr.1: 27876454-27876454 on Build GRCh38
rs1001770490 Chr.1: 27876575-27876575 on Build GRCh38
rs769663737 Chr.1: 27876595-27876595 on Build GRCh38
rs766942089 Chr.1: 27876537-27876537 on Build GRCh38
rs909910867 Chr.1: 27876523-27876523 on Build GRCh38
rs778185115 Chr.1: 27876444-27876444 on Build GRCh38
rs548173588 Chr.1: 27876490-27876490 on Build GRCh38
rs779162263 Chr.1: 27876570-27876570 on Build GRCh38
rs760603377 Chr.1: 27876480-27876480 on Build GRCh38
rs774082352 Chr.1: 27876635-27876635 on Build GRCh38
rs372097555 Chr.1: 27876631-27876631 on Build GRCh38
rs368688289 Chr.1: 27876442-27876442 on Build GRCh38
rs547985559 Chr.1: 27876583-27876583 on Build GRCh38

View Full Product Details