Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109933003-109933230 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIT2
Entrez Gene ID
9815
Gene Name
GIT ArfGAP 2
Gene Aliases
CAT-2, CAT2, PKL
UniGene
Hs.434996
Gene Chromosome Location
Chr.12: 109929792-109996389 on Build GRCh38
Transcripts
NM_001135213.1 NM_001135214.1 NM_014776.3 NM_057169.3 NM_057170.3 XM_005253997.3 XM_005254000.3 XM_006719707.3 XM_006719708.3 XM_006719709.3 XM_006719712.3 XM_017020258.1 XM_017020259.1 XM_017020260.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6671101 c.2216C>T p.A739V Chr.12: 109933042-109933042 on Build GRCh38
934887 c.2226C>T p.I742I Chr.12: 109933032-109933032 on Build GRCh38
6671094 c.2227G>A p.A743T Chr.12: 109933031-109933031 on Build GRCh38
1208204 c.2159G>T p.G720V Chr.12: 109933099-109933099 on Build GRCh38
7417260 c.2236G>A p.A746T Chr.12: 109933022-109933022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778688948 Chr.12: 109933023-109933023 on Build GRCh38
rs138906145 Chr.12: 109933041-109933041 on Build GRCh38
rs772784664 Chr.12: 109933022-109933022 on Build GRCh38
rs780219011 Chr.12: 109933072-109933072 on Build GRCh38
rs1018959710 Chr.12: 109933188-109933188 on Build GRCh38
rs779285843 Chr.12: 109933045-109933045 on Build GRCh38
rs994663834 Chr.12: 109933134-109933134 on Build GRCh38
rs753666949 Chr.12: 109933156-109933156 on Build GRCh38
rs752619910 Chr.12: 109933132-109933132 on Build GRCh38
rs748614133 Chr.12: 109933168-109933168 on Build GRCh38
rs754956489 Chr.12: 109933164-109933164 on Build GRCh38
rs778158210 Chr.12: 109933185-109933185 on Build GRCh38
rs748515904 Chr.12: 109933091-109933091 on Build GRCh38
rs772872967 Chr.12: 109933097-109933097 on Build GRCh38
rs201742073 Chr.12: 109933076-109933076 on Build GRCh38
rs770460324 Chr.12: 109933108-109933108 on Build GRCh38
rs770255193 Chr.12: 109933173-109933173 on Build GRCh38
rs780137647 Chr.12: 109933157-109933157 on Build GRCh38
rs755918306 Chr.12: 109933043-109933043 on Build GRCh38
rs750638470 Chr.12: 109933062-109933062 on Build GRCh38
rs186643308 Chr.12: 109933141-109933141 on Build GRCh38
rs1046536690 Chr.12: 109933137-109933137 on Build GRCh38
rs761498479 Chr.12: 109933199-109933200 on Build GRCh38
rs758667543 Chr.12: 109933063-109933063 on Build GRCh38
rs752551743 Chr.12: 109933165-109933165 on Build GRCh38
rs781680493 Chr.12: 109933167-109933167 on Build GRCh38
rs765864824 Chr.12: 109933025-109933025 on Build GRCh38
rs781492736 Chr.12: 109933087-109933087 on Build GRCh38
rs545428499 Chr.12: 109933202-109933202 on Build GRCh38
rs894926967 Chr.12: 109933126-109933126 on Build GRCh38
rs181939506 Chr.12: 109933077-109933077 on Build GRCh38
rs149426484 Chr.12: 109933092-109933092 on Build GRCh38
rs767491436 Chr.12: 109933122-109933122 on Build GRCh38
rs751666546 Chr.12: 109933161-109933161 on Build GRCh38
rs372439653 Chr.12: 109933104-109933104 on Build GRCh38
rs751012181 Chr.12: 109933031-109933031 on Build GRCh38
rs1012001100 Chr.12: 109933136-109933136 on Build GRCh38
rs764025212 Chr.12: 109933146-109933146 on Build GRCh38
rs758962325 Chr.12: 109933114-109933114 on Build GRCh38
rs150244358 Chr.12: 109933038-109933038 on Build GRCh38
rs557036414 Chr.12: 109933110-109933110 on Build GRCh38
rs759559369 Chr.12: 109933032-109933032 on Build GRCh38
rs907384183 Chr.12: 109933147-109933147 on Build GRCh38
rs762643976 Chr.12: 109933100-109933100 on Build GRCh38

View Full Product Details