Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 69255777-69256032 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLCE
Entrez Gene ID
26035
Gene Name
glucuronic acid epimerase
Gene Aliases
HSEPI
UniGene
Hs.183006
Gene Chromosome Location
Chr.15: 69160555-69272209 on Build GRCh38
Transcripts
NM_001324091.1 NM_001324092.1 NM_001324093.1 NM_001324094.1 NM_015554.2 XM_005254298.3 XM_017022073.1 XM_017022074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6671408 c.125G>A p.R42H Chr.15: 69255931-69255931 on Build GRCh38
6142878 c.178G>C p.E60Q Chr.15: 69255984-69255984 on Build GRCh38
1238987 c.51C>A p.C17* Chr.15: 69255857-69255857 on Build GRCh38
6246887 c.148G>T p.D50Y Chr.15: 69255954-69255954 on Build GRCh38
3387013 c.19C>A p.R7R Chr.15: 69255825-69255825 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771586419 Chr.15: 69255969-69255969 on Build GRCh38
rs150155724 Chr.15: 69255857-69255857 on Build GRCh38
rs892803570 Chr.15: 69255901-69255901 on Build GRCh38
rs201399708 Chr.15: 69255927-69255927 on Build GRCh38
rs1039333309 Chr.15: 69255896-69255896 on Build GRCh38
rs777623684 Chr.15: 69255931-69255931 on Build GRCh38
rs146925203 Chr.15: 69255940-69255940 on Build GRCh38
rs548737627 Chr.15: 69255887-69255887 on Build GRCh38
rs748092450 Chr.15: 69255814-69255814 on Build GRCh38
rs878960467 Chr.15: 69255974-69255974 on Build GRCh38
rs12440300 Chr.15: 69255999-69255999 on Build GRCh38
rs765792674 Chr.15: 69255868-69255868 on Build GRCh38
rs770428454 Chr.15: 69255994-69255994 on Build GRCh38
rs774052098 Chr.15: 69255832-69255832 on Build GRCh38
rs745504393 Chr.15: 69255964-69255964 on Build GRCh38
rs769614046 Chr.15: 69255820-69255820 on Build GRCh38
rs773019085 Chr.15: 69255821-69255821 on Build GRCh38
rs766577129 Chr.15: 69255885-69255885 on Build GRCh38
rs758767598 Chr.15: 69255882-69255882 on Build GRCh38
rs773925648 Chr.15: 69255960-69255960 on Build GRCh38
rs890910735 Chr.15: 69255869-69255869 on Build GRCh38
rs770703255 Chr.15: 69255935-69255935 on Build GRCh38
rs552257481 Chr.15: 69255810-69255810 on Build GRCh38
rs762441867 Chr.15: 69255860-69255860 on Build GRCh38
rs375636790 Chr.15: 69255878-69255878 on Build GRCh38
rs755058720 Chr.15: 69255892-69255892 on Build GRCh38
rs762181628 Chr.15: 69255924-69255924 on Build GRCh38
rs749027018 Chr.15: 69255825-69255825 on Build GRCh38
rs184777253 Chr.15: 69256008-69256008 on Build GRCh38
rs140830366 Chr.15: 69255811-69255811 on Build GRCh38
rs1044389862 Chr.15: 69255984-69255984 on Build GRCh38
rs749152412 Chr.15: 69255934-69255934 on Build GRCh38
rs756061835 Chr.15: 69255928-69255928 on Build GRCh38
rs1017417378 Chr.15: 69255866-69255866 on Build GRCh38
rs569294450 Chr.15: 69255835-69255835 on Build GRCh38
rs193005521 Chr.15: 69256007-69256007 on Build GRCh38
rs770578250 Chr.15: 69255827-69255827 on Build GRCh38
rs760203085 Chr.15: 69255973-69255973 on Build GRCh38
rs866903885 Chr.15: 69255879-69255879 on Build GRCh38
rs751765489 Chr.15: 69256011-69256011 on Build GRCh38
rs138637994 Chr.15: 69255858-69255858 on Build GRCh38
rs775089643 Chr.15: 69255970-69255970 on Build GRCh38

View Full Product Details