Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6445215-6445486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD27
Entrez Gene ID
939
Gene Name
CD27 molecule
Gene Aliases
S152, S152. LPFS2, T14, TNFRSF7, Tp55
UniGene
Hs.355307
Gene Chromosome Location
Chr.12: 6444867-6451718 on Build GRCh38
Transcripts
NM_001242.4 XM_017020232.1 XM_017020234.1 NR_015382.2

Gene Symbol
CD27-AS1
Entrez Gene ID
678655
Gene Name
CD27 antisense RNA 1
Gene Aliases
-
UniGene
Hs.655224
Gene Chromosome Location
Chr.12: 6439001-6451517 on Build GRCh38
Transcripts
NM_001242.4 XM_017020232.1 XM_017020234.1 NR_015382.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3736072 c.175G>A p.A59T Chr.12: 6445462-6445462 on Build GRCh38
4670433 c.181_182delCA p.Q61fs*2 Chr.12: 6445468-6445469 on Build GRCh38
1363666 c.148G>A p.V50M Chr.12: 6445435-6445435 on Build GRCh38
468777 c.179C>A p.A60D Chr.12: 6445466-6445466 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756940583 Chr.12: 6445451-6445451 on Build GRCh38
rs763617266 Chr.12: 6445435-6445435 on Build GRCh38
rs753465972 Chr.12: 6445241-6445241 on Build GRCh38
rs372790118 Chr.12: 6445267-6445267 on Build GRCh38
rs529096536 Chr.12: 6445463-6445463 on Build GRCh38
rs747125906 Chr.12: 6445237-6445238 on Build GRCh38
rs370603141 Chr.12: 6445419-6445419 on Build GRCh38
rs1051544497 Chr.12: 6445396-6445396 on Build GRCh38
rs377143297 Chr.12: 6445268-6445268 on Build GRCh38
rs759236596 Chr.12: 6445415-6445415 on Build GRCh38
rs769967539 Chr.12: 6445270-6445270 on Build GRCh38
rs982628572 Chr.12: 6445260-6445260 on Build GRCh38
rs200745064 Chr.12: 6445454-6445454 on Build GRCh38
rs997270807 Chr.12: 6445387-6445387 on Build GRCh38
rs376133215 Chr.12: 6445246-6445246 on Build GRCh38
rs144208131 Chr.12: 6445293-6445293 on Build GRCh38
rs199970363 Chr.12: 6445272-6445272 on Build GRCh38
rs760226591 Chr.12: 6445434-6445434 on Build GRCh38
rs919567116 Chr.12: 6445386-6445386 on Build GRCh38
rs949488792 Chr.12: 6445276-6445276 on Build GRCh38
rs770989481 Chr.12: 6445382-6445382 on Build GRCh38
rs753305875 Chr.12: 6445442-6445442 on Build GRCh38
rs866081455 Chr.12: 6445364-6445364 on Build GRCh38
rs781593353 Chr.12: 6445440-6445440 on Build GRCh38
rs71584821 Chr.12: 6445305-6445305 on Build GRCh38
rs747271704 Chr.12: 6445259-6445259 on Build GRCh38
rs201916358 Chr.12: 6445453-6445453 on Build GRCh38
rs767508630 Chr.12: 6445416-6445416 on Build GRCh38
rs763774879 Chr.12: 6445236-6445236 on Build GRCh38
rs573505026 Chr.12: 6445252-6445252 on Build GRCh38
rs2534719 Chr.12: 6445263-6445263 on Build GRCh38
rs749551392 Chr.12: 6445374-6445374 on Build GRCh38
rs938255517 Chr.12: 6445349-6445349 on Build GRCh38
rs901358851 Chr.12: 6445375-6445375 on Build GRCh38
rs768803245 Chr.12: 6445411-6445415 on Build GRCh38
rs73054312 Chr.12: 6445304-6445304 on Build GRCh38
rs774207253 Chr.12: 6445414-6445414 on Build GRCh38
rs397514667 Chr.12: 6445445-6445445 on Build GRCh38
rs25680 Chr.12: 6445462-6445462 on Build GRCh38

View Full Product Details