Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 22600976-22601245 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf58
Entrez Gene ID
541565
Gene Name
chromosome 8 open reading frame 58
Gene Aliases
-
UniGene
Hs.743508
Gene Chromosome Location
Chr.8: 22599601-22604150 on Build GRCh38
Transcripts
NM_001013842.2 NM_001198827.1 NM_173686.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5440980 c.287C>G p.S96* Chr.8: 22601128-22601128 on Build GRCh38
6392439 c.359G>A p.R120Q Chr.8: 22601200-22601200 on Build GRCh38
1098125 c.365G>T p.R122L Chr.8: 22601206-22601206 on Build GRCh38
2786282 c.165C>T p.V55V Chr.8: 22601006-22601006 on Build GRCh38
4473725 c.186C>T p.L62L Chr.8: 22601027-22601027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769069083 Chr.8: 22601111-22601111 on Build GRCh38
rs141287905 Chr.8: 22601208-22601208 on Build GRCh38
rs1030676327 Chr.8: 22601109-22601109 on Build GRCh38
rs774674193 Chr.8: 22601223-22601223 on Build GRCh38
rs750265629 Chr.8: 22601176-22601176 on Build GRCh38
rs1050988814 Chr.8: 22601187-22601187 on Build GRCh38
rs766192741 Chr.8: 22601048-22601048 on Build GRCh38
rs768803044 Chr.8: 22601057-22601057 on Build GRCh38
rs138371466 Chr.8: 22601006-22601006 on Build GRCh38
rs561945277 Chr.8: 22601091-22601091 on Build GRCh38
rs750822224 Chr.8: 22601003-22601003 on Build GRCh38
rs749235252 Chr.8: 22601033-22601033 on Build GRCh38
rs755822102 Chr.8: 22601088-22601088 on Build GRCh38
rs763954294 Chr.8: 22601110-22601110 on Build GRCh38
rs772012529 Chr.8: 22601041-22601041 on Build GRCh38
rs916738772 Chr.8: 22601128-22601128 on Build GRCh38
rs868666321 Chr.8: 22601119-22601119 on Build GRCh38
rs773605548 Chr.8: 22601065-22601065 on Build GRCh38
rs753548607 Chr.8: 22601162-22601162 on Build GRCh38
rs370995356 Chr.8: 22601000-22601000 on Build GRCh38
rs747235215 Chr.8: 22601055-22601055 on Build GRCh38
rs940786741 Chr.8: 22601155-22601155 on Build GRCh38
rs763179620 Chr.8: 22601077-22601077 on Build GRCh38
rs754660803 Chr.8: 22601017-22601017 on Build GRCh38
rs778663501 Chr.8: 22601101-22601101 on Build GRCh38
rs969986164 Chr.8: 22601131-22601131 on Build GRCh38
rs868078576 Chr.8: 22601130-22601131 on Build GRCh38
rs776911845 Chr.8: 22601059-22601059 on Build GRCh38
rs774763849 Chr.8: 22601156-22601156 on Build GRCh38
rs557683546 Chr.8: 22601178-22601178 on Build GRCh38
rs368897134 Chr.8: 22601040-22601040 on Build GRCh38
rs778491158 Chr.8: 22601085-22601085 on Build GRCh38
rs769696478 Chr.8: 22601206-22601206 on Build GRCh38
rs374180743 Chr.8: 22601016-22601016 on Build GRCh38
rs764804889 Chr.8: 22601171-22601171 on Build GRCh38
rs534172908 Chr.8: 22601180-22601180 on Build GRCh38
rs771467426 Chr.8: 22601217-22601217 on Build GRCh38
rs757107894 Chr.8: 22601141-22601142 on Build GRCh38
rs529077410 Chr.8: 22601108-22601108 on Build GRCh38
rs751444263 Chr.8: 22601184-22601184 on Build GRCh38
rs543335279 Chr.8: 22601082-22601082 on Build GRCh38
rs777278509 Chr.8: 22601019-22601019 on Build GRCh38
rs765705925 Chr.8: 22601064-22601064 on Build GRCh38
rs1007169902 Chr.8: 22601209-22601209 on Build GRCh38
rs559034053 Chr.8: 22601147-22601147 on Build GRCh38
rs370688473 Chr.8: 22601142-22601142 on Build GRCh38
rs757207422 Chr.8: 22601038-22601038 on Build GRCh38
rs201978138 Chr.8: 22601001-22601001 on Build GRCh38
rs201976769 Chr.8: 22601036-22601036 on Build GRCh38
rs775810670 Chr.8: 22601046-22601046 on Build GRCh38
rs769941496 Chr.8: 22601146-22601146 on Build GRCh38
rs199978994 Chr.8: 22601110-22601110 on Build GRCh38
rs752369873 Chr.8: 22601014-22601014 on Build GRCh38
rs992425465 Chr.8: 22601117-22601117 on Build GRCh38
rs749791714 Chr.8: 22601210-22601210 on Build GRCh38
rs374640891 Chr.8: 22601150-22601150 on Build GRCh38
rs757194612 Chr.8: 22601093-22601093 on Build GRCh38
rs1033716854 Chr.8: 22601116-22601116 on Build GRCh38
rs368626009 Chr.8: 22601205-22601205 on Build GRCh38
rs765617757 Chr.8: 22601002-22601002 on Build GRCh38
rs35678720 Chr.8: 22601049-22601049 on Build GRCh38
rs752279989 Chr.8: 22601084-22601084 on Build GRCh38
rs778735614 Chr.8: 22601039-22601039 on Build GRCh38
rs767208210 Chr.8: 22601007-22601007 on Build GRCh38
rs760287999 Chr.8: 22601161-22601161 on Build GRCh38
rs550984232 Chr.8: 22601199-22601199 on Build GRCh38
rs998075395 Chr.8: 22601004-22601004 on Build GRCh38
rs781224538 Chr.8: 22601200-22601200 on Build GRCh38
rs780932501 Chr.8: 22601209-22601210 on Build GRCh38
rs761185984 Chr.8: 22601165-22601165 on Build GRCh38
rs771271445 Chr.8: 22601148-22601148 on Build GRCh38
rs879214142 Chr.8: 22601141-22601141 on Build GRCh38
rs781303712 Chr.8: 22601129-22601129 on Build GRCh38
rs751504768 Chr.8: 22601142-22601142 on Build GRCh38
rs376060539 Chr.8: 22601211-22601211 on Build GRCh38
rs750402120 Chr.8: 22601104-22601104 on Build GRCh38
rs981519326 Chr.8: 22601204-22601204 on Build GRCh38
rs11552429 Chr.8: 22601092-22601092 on Build GRCh38

View Full Product Details