Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3668858-3669113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM33
Entrez Gene ID
80332
Gene Name
ADAM metallopeptidase domain 33
Gene Aliases
C20orf153, DJ964F7.1
UniGene
Hs.173716
Gene Chromosome Location
Chr.20: 3667973-3682131 on Build GRCh38
Transcripts
NM_001282447.2 NM_025220.4 NM_153202.3 XM_005260843.1 XM_006723639.1 XM_006723640.1 XM_011529366.1 XM_011529367.1 XM_011529368.1 XM_011529373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7123276 c.2414T>G p.V805G Chr.20: 3668991-3668991 on Build GRCh38
1729281 c.2417A>T p.Q806L Chr.20: 3668988-3668988 on Build GRCh38
3546062 c.2405C>A p.A802E Chr.20: 3669000-3669000 on Build GRCh38
4421064 c.2405-9C>A p.? Chr.20: 3669009-3669009 on Build GRCh38
4967852 c.2409_2421>TCATG p.Q804fs*>8 Chr.20: 3668984-3668996 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760772020 Chr.20: 3669047-3669047 on Build GRCh38
rs111467497 Chr.20: 3668919-3668920 on Build GRCh38
rs543749 Chr.20: 3669032-3669032 on Build GRCh38
rs768485706 Chr.20: 3669043-3669043 on Build GRCh38
rs867216249 Chr.20: 3669033-3669033 on Build GRCh38
rs748496134 Chr.20: 3668917-3668917 on Build GRCh38
rs376802146 Chr.20: 3669026-3669026 on Build GRCh38
rs932335729 Chr.20: 3669062-3669062 on Build GRCh38
rs769683039 Chr.20: 3669037-3669037 on Build GRCh38
rs767094027 Chr.20: 3669015-3669017 on Build GRCh38
rs765444719 Chr.20: 3669088-3669088 on Build GRCh38
rs779264848 Chr.20: 3668998-3668998 on Build GRCh38
rs628965 Chr.20: 3669066-3669066 on Build GRCh38
rs773225118 Chr.20: 3669046-3669046 on Build GRCh38
rs746532303 Chr.20: 3669027-3669027 on Build GRCh38
rs376742291 Chr.20: 3668930-3668930 on Build GRCh38
rs766607508 Chr.20: 3668992-3668992 on Build GRCh38
rs772984536 Chr.20: 3668927-3668927 on Build GRCh38
rs55798864 Chr.20: 3668887-3668887 on Build GRCh38
rs200145332 Chr.20: 3668989-3668989 on Build GRCh38
rs369852454 Chr.20: 3669031-3669031 on Build GRCh38
rs369283635 Chr.20: 3668943-3668943 on Build GRCh38
rs763016968 Chr.20: 3669040-3669040 on Build GRCh38
rs970050517 Chr.20: 3668879-3668879 on Build GRCh38
rs745620382 Chr.20: 3669035-3669035 on Build GRCh38
rs756268292 Chr.20: 3668990-3668990 on Build GRCh38
rs537914783 Chr.20: 3668901-3668901 on Build GRCh38
rs749663083 Chr.20: 3668916-3668916 on Build GRCh38
rs757840545 Chr.20: 3669014-3669014 on Build GRCh38
rs202190276 Chr.20: 3668953-3668953 on Build GRCh38
rs765046256 Chr.20: 3668952-3668952 on Build GRCh38
rs977556963 Chr.20: 3668922-3668922 on Build GRCh38
rs747580126 Chr.20: 3668919-3668919 on Build GRCh38
rs17548837 Chr.20: 3668987-3668994 on Build GRCh38
rs142167062 Chr.20: 3668972-3668972 on Build GRCh38
rs920888833 Chr.20: 3669057-3669057 on Build GRCh38
rs541517124 Chr.20: 3668881-3668881 on Build GRCh38
rs137919189 Chr.20: 3668971-3668971 on Build GRCh38
rs628977 Chr.20: 3669074-3669074 on Build GRCh38
rs777350843 Chr.20: 3669020-3669020 on Build GRCh38
rs371393373 Chr.20: 3669008-3669008 on Build GRCh38
rs3918401 Chr.20: 3668993-3668993 on Build GRCh38
rs1048308883 Chr.20: 3669041-3669041 on Build GRCh38
rs201565525 Chr.20: 3668995-3668995 on Build GRCh38
rs146576636 Chr.20: 3668986-3668993 on Build GRCh38
rs550126514 Chr.20: 3668931-3668931 on Build GRCh38
rs549135797 Chr.20: 3668883-3668883 on Build GRCh38
rs386811870 Chr.20: 3669066-3669074 on Build GRCh38
rs1048080014 Chr.20: 3669064-3669065 on Build GRCh38
rs775280544 Chr.20: 3669038-3669038 on Build GRCh38

View Full Product Details