Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46016565-46016832 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC61
Entrez Gene ID
729440
Gene Name
coiled-coil domain containing 61
Gene Aliases
-
UniGene
Hs.515479
Gene Chromosome Location
Chr.19: 45995457-46018616 on Build GRCh38
Transcripts
NM_001267723.1 XM_005259191.3 XM_005259192.4 XM_011527257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6427025 c.1241C>A p.T414N Chr.19: 46016786-46016786 on Build GRCh38
6415407 c.1262A>G p.N421S Chr.19: 46016807-46016807 on Build GRCh38
7230580 c.1197G>T p.P399P Chr.19: 46016742-46016742 on Build GRCh38
3692894 c.1214G>T p.R405L Chr.19: 46016759-46016759 on Build GRCh38
6252250 c.1234G>T p.A412S Chr.19: 46016779-46016779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975630905 Chr.19: 46016810-46016810 on Build GRCh38
rs773355967 Chr.19: 46016756-46016756 on Build GRCh38
rs772312800 Chr.19: 46016694-46016696 on Build GRCh38
rs971332433 Chr.19: 46016674-46016674 on Build GRCh38
rs754371850 Chr.19: 46016661-46016661 on Build GRCh38
rs2287306 Chr.19: 46016768-46016768 on Build GRCh38
rs747275588 Chr.19: 46016658-46016658 on Build GRCh38
rs760544095 Chr.19: 46016711-46016711 on Build GRCh38
rs769836334 Chr.19: 46016678-46016678 on Build GRCh38
rs764869678 Chr.19: 46016726-46016726 on Build GRCh38
rs199634925 Chr.19: 46016699-46016699 on Build GRCh38
rs773130238 Chr.19: 46016682-46016682 on Build GRCh38
rs772116307 Chr.19: 46016763-46016763 on Build GRCh38
rs775272377 Chr.19: 46016688-46016688 on Build GRCh38
rs755316027 Chr.19: 46016741-46016741 on Build GRCh38
rs1017230484 Chr.19: 46016799-46016799 on Build GRCh38
rs761579264 Chr.19: 46016809-46016809 on Build GRCh38
rs1013610449 Chr.19: 46016693-46016693 on Build GRCh38
rs763949486 Chr.19: 46016783-46016783 on Build GRCh38
rs370235806 Chr.19: 46016691-46016691 on Build GRCh38
rs748266892 Chr.19: 46016678-46016681 on Build GRCh38
rs1026391943 Chr.19: 46016702-46016702 on Build GRCh38
rs776525592 Chr.19: 46016792-46016792 on Build GRCh38
rs769958653 Chr.19: 46016749-46016749 on Build GRCh38
rs763896331 Chr.19: 46016712-46016712 on Build GRCh38
rs955223024 Chr.19: 46016814-46016814 on Build GRCh38
rs760593504 Chr.19: 46016775-46016775 on Build GRCh38
rs373742598 Chr.19: 46016722-46016722 on Build GRCh38
rs776826413 Chr.19: 46016671-46016671 on Build GRCh38
rs144277506 Chr.19: 46016811-46016811 on Build GRCh38
rs1002753960 Chr.19: 46016616-46016616 on Build GRCh38
rs950717279 Chr.19: 46016761-46016761 on Build GRCh38
rs530327465 Chr.19: 46016615-46016615 on Build GRCh38
rs780486515 Chr.19: 46016655-46016655 on Build GRCh38
rs550071010 Chr.19: 46016673-46016673 on Build GRCh38
rs749260599 Chr.19: 46016758-46016758 on Build GRCh38
rs1012794725 Chr.19: 46016595-46016595 on Build GRCh38
rs774660406 Chr.19: 46016662-46016663 on Build GRCh38
rs753516618 Chr.19: 46016720-46016720 on Build GRCh38
rs749850932 Chr.19: 46016737-46016737 on Build GRCh38
rs781435168 Chr.19: 46016742-46016742 on Build GRCh38
rs1022870134 Chr.19: 46016639-46016639 on Build GRCh38
rs748403747 Chr.19: 46016745-46016745 on Build GRCh38
rs775361811 Chr.19: 46016705-46016705 on Build GRCh38
rs780355466 Chr.19: 46016740-46016740 on Build GRCh38

View Full Product Details