Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67830327-67830588 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPT
Entrez Gene ID
80152
Gene Name
centromere protein T
Gene Aliases
C16orf56, CENP-T
UniGene
Hs.288382
Gene Chromosome Location
Chr.16: 67828157-67847458 on Build GRCh38
Transcripts
NM_025082.3 XM_006721277.2 XM_011523344.1 XM_011523345.1 XM_017023712.1 XM_017023713.1 XM_017023714.1 XM_017023715.1 XM_017023716.1 XM_017023717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4432866 c.764A>G p.Y255C Chr.16: 67830488-67830488 on Build GRCh38
5385194 c.745G>A p.V249I Chr.16: 67830507-67830507 on Build GRCh38
6626693 c.862+4A>C p.? Chr.16: 67830386-67830386 on Build GRCh38
4942758 c.854A>G p.Q285R Chr.16: 67830398-67830398 on Build GRCh38
6522948 c.793G>T p.G265W Chr.16: 67830459-67830459 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773369608 Chr.16: 67830432-67830432 on Build GRCh38
rs749801113 Chr.16: 67830381-67830381 on Build GRCh38
rs755155899 Chr.16: 67830460-67830460 on Build GRCh38
rs920467791 Chr.16: 67830405-67830405 on Build GRCh38
rs944390362 Chr.16: 67830450-67830450 on Build GRCh38
rs762220056 Chr.16: 67830429-67830429 on Build GRCh38
rs200402335 Chr.16: 67830367-67830367 on Build GRCh38
rs771671855 Chr.16: 67830556-67830556 on Build GRCh38
rs778175741 Chr.16: 67830497-67830497 on Build GRCh38
rs749885312 Chr.16: 67830469-67830469 on Build GRCh38
rs769575578 Chr.16: 67830447-67830447 on Build GRCh38
rs199963870 Chr.16: 67830523-67830523 on Build GRCh38
rs754356295 Chr.16: 67830489-67830489 on Build GRCh38
rs747138104 Chr.16: 67830433-67830433 on Build GRCh38
rs774071761 Chr.16: 67830544-67830544 on Build GRCh38
rs747232781 Chr.16: 67830520-67830520 on Build GRCh38
rs188345411 Chr.16: 67830463-67830463 on Build GRCh38
rs117141770 Chr.16: 67830492-67830492 on Build GRCh38
rs779478818 Chr.16: 67830427-67830427 on Build GRCh38
rs1035684401 Chr.16: 67830532-67830532 on Build GRCh38
rs575604055 Chr.16: 67830470-67830470 on Build GRCh38
rs72790365 Chr.16: 67830428-67830428 on Build GRCh38
rs774198099 Chr.16: 67830461-67830461 on Build GRCh38
rs540685212 Chr.16: 67830494-67830494 on Build GRCh38
rs186354372 Chr.16: 67830545-67830545 on Build GRCh38
rs202014360 Chr.16: 67830471-67830471 on Build GRCh38
rs951033959 Chr.16: 67830490-67830490 on Build GRCh38
rs998127588 Chr.16: 67830481-67830481 on Build GRCh38
rs755350013 Chr.16: 67830407-67830407 on Build GRCh38
rs762843667 Chr.16: 67830457-67830457 on Build GRCh38
rs56045040 Chr.16: 67830493-67830493 on Build GRCh38
rs768583120 Chr.16: 67830531-67830531 on Build GRCh38
rs772898942 Chr.16: 67830561-67830561 on Build GRCh38
rs201152976 Chr.16: 67830447-67830447 on Build GRCh38
rs368284034 Chr.16: 67830526-67830526 on Build GRCh38
rs375134489 Chr.16: 67830563-67830563 on Build GRCh38
rs780631856 Chr.16: 67830375-67830375 on Build GRCh38
rs372208707 Chr.16: 67830448-67830448 on Build GRCh38
rs768481051 Chr.16: 67830458-67830458 on Build GRCh38
rs767009908 Chr.16: 67830365-67830365 on Build GRCh38
rs756958493 Chr.16: 67830354-67830354 on Build GRCh38
rs781183528 Chr.16: 67830524-67830524 on Build GRCh38

View Full Product Details