Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30112871-30113144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD3
Entrez Gene ID
79153
Gene Name
glycerophosphodiester phosphodiesterase domain containing 3
Gene Aliases
GDE7
UniGene
Hs.289015
Gene Chromosome Location
Chr.16: 30104810-30113557 on Build GRCh38
Transcripts
NM_024307.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4950171 c.154C>G p.L52V Chr.16: 30113050-30113050 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759377513 Chr.16: 30112973-30112973 on Build GRCh38
rs368935847 Chr.16: 30113039-30113039 on Build GRCh38
rs555397157 Chr.16: 30113110-30113110 on Build GRCh38
rs771883070 Chr.16: 30113102-30113102 on Build GRCh38
rs201475067 Chr.16: 30113074-30113074 on Build GRCh38
rs187844693 Chr.16: 30113042-30113042 on Build GRCh38
rs1020406973 Chr.16: 30112900-30112900 on Build GRCh38
rs989811316 Chr.16: 30112896-30112896 on Build GRCh38
rs769645432 Chr.16: 30113121-30113121 on Build GRCh38
rs937843028 Chr.16: 30112987-30112987 on Build GRCh38
rs750822886 Chr.16: 30113006-30113006 on Build GRCh38
rs548990032 Chr.16: 30113071-30113071 on Build GRCh38
rs927397949 Chr.16: 30112955-30112956 on Build GRCh38
rs758699537 Chr.16: 30113015-30113015 on Build GRCh38
rs967143615 Chr.16: 30112945-30112945 on Build GRCh38
rs34176126 Chr.16: 30112912-30112913 on Build GRCh38
rs960836995 Chr.16: 30113017-30113017 on Build GRCh38
rs981540280 Chr.16: 30112906-30112906 on Build GRCh38
rs755824594 Chr.16: 30113023-30113024 on Build GRCh38
rs183129319 Chr.16: 30112990-30112990 on Build GRCh38
rs775004990 Chr.16: 30113107-30113107 on Build GRCh38
rs781462726 Chr.16: 30112972-30112972 on Build GRCh38
rs907900868 Chr.16: 30113075-30113075 on Build GRCh38
rs763744371 Chr.16: 30113113-30113113 on Build GRCh38
rs774011500 Chr.16: 30113088-30113088 on Build GRCh38
rs373056992 Chr.16: 30113079-30113079 on Build GRCh38
rs565937012 Chr.16: 30112917-30112917 on Build GRCh38
rs752119403 Chr.16: 30112985-30112985 on Build GRCh38
rs752169145 Chr.16: 30113037-30113037 on Build GRCh38
rs377658910 Chr.16: 30112956-30112957 on Build GRCh38
rs745860273 Chr.16: 30113097-30113097 on Build GRCh38
rs763688835 Chr.16: 30112991-30112991 on Build GRCh38
rs1040444922 Chr.16: 30113117-30113117 on Build GRCh38
rs981121689 Chr.16: 30112977-30112977 on Build GRCh38
rs780442950 Chr.16: 30113019-30113019 on Build GRCh38
rs748187597 Chr.16: 30113063-30113063 on Build GRCh38
rs767119474 Chr.16: 30112983-30112983 on Build GRCh38
rs1020242421 Chr.16: 30112892-30112892 on Build GRCh38

View Full Product Details