Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6269858-6270103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCKBR
Entrez Gene ID
887
Gene Name
cholecystokinin B receptor
Gene Aliases
CCK-B, CCK2R, GASR
UniGene
Hs.203
Gene Chromosome Location
Chr.11: 6259611-6272127 on Build GRCh38
Transcripts
NM_001318029.1 NM_176875.3 XM_005253210.1 XM_017018516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
304619 c.403G>T p.G135W Chr.11: 6269920-6269920 on Build GRCh38
6133972 c.418G>T p.V140L Chr.11: 6270102-6270102 on Build GRCh38
6133971 c.418G>T p.V140L Chr.11: 6270102-6270102 on Build GRCh38
5601206 c.404G>A p.G135E Chr.11: 6270088-6270088 on Build GRCh38
6133976 c.360C>T p.F120F Chr.11: 6269877-6269877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762562421 Chr.11: 6269955-6269955 on Build GRCh38
rs201899108 Chr.11: 6270060-6270060 on Build GRCh38
rs749283305 Chr.11: 6270056-6270056 on Build GRCh38
rs996748863 Chr.11: 6269996-6269996 on Build GRCh38
rs781195547 Chr.11: 6269902-6269902 on Build GRCh38
rs78670873 Chr.11: 6270078-6270078 on Build GRCh38
rs754661690 Chr.11: 6269900-6269900 on Build GRCh38
rs199500410 Chr.11: 6269924-6269924 on Build GRCh38
rs199529328 Chr.11: 6270072-6270072 on Build GRCh38
rs200605105 Chr.11: 6269938-6269938 on Build GRCh38
rs767696728 Chr.11: 6269934-6269934 on Build GRCh38
rs762025188 Chr.11: 6269888-6269888 on Build GRCh38
rs201414174 Chr.11: 6269946-6269946 on Build GRCh38
rs199733398 Chr.11: 6269948-6269948 on Build GRCh38
rs780852412 Chr.11: 6269974-6269974 on Build GRCh38
rs200524431 Chr.11: 6269904-6269904 on Build GRCh38
rs759821680 Chr.11: 6269925-6269925 on Build GRCh38
rs761050844 Chr.11: 6269942-6269942 on Build GRCh38
rs541507223 Chr.11: 6269988-6269988 on Build GRCh38
rs955339321 Chr.11: 6270017-6270017 on Build GRCh38
rs574726739 Chr.11: 6270011-6270011 on Build GRCh38
rs200916711 Chr.11: 6269957-6269957 on Build GRCh38
rs773163565 Chr.11: 6269906-6269906 on Build GRCh38
rs770942022 Chr.11: 6269916-6269916 on Build GRCh38
rs761166174 Chr.11: 6270063-6270063 on Build GRCh38
rs752470926 Chr.11: 6270039-6270039 on Build GRCh38
rs766339407 Chr.11: 6270010-6270010 on Build GRCh38
rs200867208 Chr.11: 6270047-6270047 on Build GRCh38
rs541728415 Chr.11: 6270040-6270040 on Build GRCh38
rs199737121 Chr.11: 6270022-6270022 on Build GRCh38
rs766229492 Chr.11: 6269920-6269920 on Build GRCh38
rs773303225 Chr.11: 6269963-6269963 on Build GRCh38
rs76412645 Chr.11: 6270073-6270073 on Build GRCh38
rs117657121 Chr.11: 6269997-6269997 on Build GRCh38
rs142903629 Chr.11: 6269914-6269914 on Build GRCh38
rs1805002 Chr.11: 6269890-6269890 on Build GRCh38
rs1045402791 Chr.11: 6269929-6269929 on Build GRCh38
rs755430730 Chr.11: 6270071-6270071 on Build GRCh38
rs901995492 Chr.11: 6270053-6270053 on Build GRCh38

View Full Product Details