Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 103095098-103095367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BDH2
Entrez Gene ID
56898
Gene Name
3-hydroxybutyrate dehydrogenase, type 2
Gene Aliases
DHRS6, EFA6R, PRO20933, SDR15C1, UCPA-OR, UNQ6308
UniGene
Hs.124696
Gene Chromosome Location
Chr.4: 103077618-103099867 on Build GRCh38
Transcripts
NM_020139.3 XM_005263140.3 XM_006714274.3 XM_011532127.2 XM_011532128.2 XM_017008461.1 XM_017008462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
318913 c.137T>C p.L46P Chr.4: 103095217-103095217 on Build GRCh38
309465 c.151G>T p.G51C Chr.4: 103095203-103095203 on Build GRCh38
6371992 c.107C>A p.T36K Chr.4: 103095247-103095247 on Build GRCh38
480696 c.84A>T p.R28S Chr.4: 103095270-103095270 on Build GRCh38
3660824 c.130C>G p.Q44E Chr.4: 103095224-103095224 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs759383127 Chr.4: 103095234-103095234 on Build GRCh38
rs267599967 Chr.4: 103095226-103095226 on Build GRCh38
rs779453009 Chr.4: 103095329-103095330 on Build GRCh38
rs199621816 Chr.4: 103095274-103095274 on Build GRCh38
rs766996263 Chr.4: 103095302-103095302 on Build GRCh38
rs757943264 Chr.4: 103095153-103095153 on Build GRCh38
rs756659371 Chr.4: 103095166-103095166 on Build GRCh38
rs76142480 Chr.4: 103095267-103095267 on Build GRCh38
rs763700717 Chr.4: 103095272-103095272 on Build GRCh38
rs751094740 Chr.4: 103095273-103095273 on Build GRCh38
rs201426266 Chr.4: 103095329-103095329 on Build GRCh38
rs548876188 Chr.4: 103095122-103095122 on Build GRCh38
rs755392752 Chr.4: 103095309-103095309 on Build GRCh38
rs960990195 Chr.4: 103095134-103095134 on Build GRCh38
rs769100161 Chr.4: 103095210-103095210 on Build GRCh38
rs778019559 Chr.4: 103095240-103095240 on Build GRCh38
rs889980825 Chr.4: 103095243-103095243 on Build GRCh38
rs761254947 Chr.4: 103095140-103095140 on Build GRCh38
rs902258799 Chr.4: 103095141-103095141 on Build GRCh38
rs114978654 Chr.4: 103095296-103095296 on Build GRCh38
rs990797773 Chr.4: 103095139-103095139 on Build GRCh38
rs779208716 Chr.4: 103095190-103095190 on Build GRCh38
rs754336832 Chr.4: 103095303-103095303 on Build GRCh38
rs533864956 Chr.4: 103095158-103095158 on Build GRCh38
rs184229868 Chr.4: 103095142-103095142 on Build GRCh38
rs373465763 Chr.4: 103095321-103095321 on Build GRCh38
rs749663801 Chr.4: 103095179-103095179 on Build GRCh38
rs748268979 Chr.4: 103095202-103095202 on Build GRCh38
rs779106840 Chr.4: 103095314-103095314 on Build GRCh38
rs200156996 Chr.4: 103095206-103095206 on Build GRCh38
rs572912670 Chr.4: 103095286-103095286 on Build GRCh38
rs75286745 Chr.4: 103095261-103095261 on Build GRCh38
rs199665964 Chr.4: 103095242-103095242 on Build GRCh38
rs201463191 Chr.4: 103095205-103095205 on Build GRCh38
rs373181736 Chr.4: 103095281-103095281 on Build GRCh38
rs868591621 Chr.4: 103095254-103095254 on Build GRCh38
rs376214820 Chr.4: 103095216-103095216 on Build GRCh38
rs368308105 Chr.4: 103095176-103095176 on Build GRCh38
rs115232956 Chr.4: 103095204-103095204 on Build GRCh38
rs558801739 Chr.4: 103095163-103095163 on Build GRCh38
rs755228575 Chr.4: 103095187-103095187 on Build GRCh38
rs757746539 Chr.4: 103095275-103095279 on Build GRCh38
rs1019696209 Chr.4: 103095265-103095265 on Build GRCh38
rs764886545 Chr.4: 103095241-103095241 on Build GRCh38
rs754192502 Chr.4: 103095272-103095273 on Build GRCh38
rs746994807 Chr.4: 103095207-103095207 on Build GRCh38
rs114186764 Chr.4: 103095124-103095124 on Build GRCh38

View Full Product Details