Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 47322869-47322640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDF2
Entrez Gene ID
2658
Gene Name
growth differentiation factor 2
Gene Aliases
BMP-9, BMP9, HHT5
UniGene
Hs.279463
Gene Chromosome Location
Chr.10: 47322490-47326270 on Build GRCh38
Transcripts
NM_016204.2

View Full Product Details