Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185459736-185459985 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC110
Entrez Gene ID
256309
Gene Name
coiled-coil domain containing 110
Gene Aliases
CT52, KM-HN-1, KMHN1
UniGene
Hs.41101
Gene Chromosome Location
Chr.4: 185445182-185471759 on Build GRCh38
Transcripts
NM_001145411.1 NM_152775.3 XM_005262891.3 XM_006714172.2 XM_011531827.2 XM_011531828.2 XM_011531829.2 XM_017007987.1 XM_017007988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5400946 c.835G>A p.G279S Chr.4: 185459752-185459752 on Build GRCh38
3760523 c.626C>A p.P209Q Chr.4: 185459961-185459961 on Build GRCh38
4123747 c.741A>C p.Q247H Chr.4: 185459846-185459846 on Build GRCh38
5417425 c.813T>A p.T271T Chr.4: 185459774-185459774 on Build GRCh38
6451670 c.849G>A p.M283I Chr.4: 185459738-185459738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373736344 Chr.4: 185459921-185459921 on Build GRCh38
rs756247933 Chr.4: 185459866-185459866 on Build GRCh38
rs576367798 Chr.4: 185459791-185459791 on Build GRCh38
rs760630075 Chr.4: 185459808-185459808 on Build GRCh38
rs751679746 Chr.4: 185459836-185459836 on Build GRCh38
rs77987360 Chr.4: 185459889-185459889 on Build GRCh38
rs755221845 Chr.4: 185459845-185459845 on Build GRCh38
rs139707432 Chr.4: 185459794-185459794 on Build GRCh38
rs377132008 Chr.4: 185459947-185459947 on Build GRCh38
rs146006186 Chr.4: 185459949-185459949 on Build GRCh38
rs34846227 Chr.4: 185459815-185459816 on Build GRCh38
rs199648674 Chr.4: 185459899-185459899 on Build GRCh38
rs535673017 Chr.4: 185459834-185459834 on Build GRCh38
rs775172418 Chr.4: 185459806-185459806 on Build GRCh38
rs776696993 Chr.4: 185459822-185459822 on Build GRCh38
rs752790898 Chr.4: 185459853-185459853 on Build GRCh38
rs7699687 Chr.4: 185459961-185459961 on Build GRCh38
rs767521105 Chr.4: 185459848-185459848 on Build GRCh38
rs529227235 Chr.4: 185459826-185459826 on Build GRCh38
rs948684901 Chr.4: 185459919-185459919 on Build GRCh38
rs147398791 Chr.4: 185459771-185459771 on Build GRCh38
rs776406319 Chr.4: 185459906-185459906 on Build GRCh38
rs749842341 Chr.4: 185459871-185459871 on Build GRCh38
rs773844229 Chr.4: 185459767-185459767 on Build GRCh38
rs143728634 Chr.4: 185459868-185459868 on Build GRCh38
rs541296781 Chr.4: 185459924-185459924 on Build GRCh38
rs745480880 Chr.4: 185459892-185459892 on Build GRCh38
rs370268336 Chr.4: 185459809-185459809 on Build GRCh38
rs754595236 Chr.4: 185459829-185459829 on Build GRCh38
rs779359701 Chr.4: 185459891-185459891 on Build GRCh38
rs373953734 Chr.4: 185459896-185459896 on Build GRCh38
rs767715417 Chr.4: 185459941-185459941 on Build GRCh38
rs759699138 Chr.4: 185459928-185459928 on Build GRCh38
rs761829432 Chr.4: 185459909-185459909 on Build GRCh38
rs148706677 Chr.4: 185459905-185459905 on Build GRCh38
rs142282326 Chr.4: 185459951-185459951 on Build GRCh38

View Full Product Details