Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 24595018-24595265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP21
Entrez Gene ID
57584
Gene Name
Rho GTPase activating protein 21
Gene Aliases
ARHGAP10
UniGene
Hs.524195
Gene Chromosome Location
Chr.10: 24583609-24725421 on Build GRCh38
Transcripts
NM_020824.3 XM_005252542.3 XM_005252544.4 XM_011519602.2 XM_011519603.2 XM_011519604.2 XM_011519605.2 XM_011519606.2 XM_011519607.2 XM_017016459.1 XM_017016460.1 XM_017016461.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6394475 c.3160C>T p.P1054S Chr.10: 24595027-24595027 on Build GRCh38
5477195 c.3775A>G p.K1259E Chr.10: 24595125-24595125 on Build GRCh38
400433 c.3733G>A p.A1245T Chr.10: 24595167-24595167 on Build GRCh38
5625044 c.3112G>A p.D1038N Chr.10: 24595152-24595152 on Build GRCh38
1128078 c.3752C>G p.P1251R Chr.10: 24595148-24595148 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769101564 Chr.10: 24595146-24595146 on Build GRCh38
rs752300624 Chr.10: 24595195-24595195 on Build GRCh38
rs765846457 Chr.10: 24595048-24595048 on Build GRCh38
rs768428692 Chr.10: 24595129-24595129 on Build GRCh38
rs1030986588 Chr.10: 24595220-24595220 on Build GRCh38
rs762321576 Chr.10: 24595151-24595151 on Build GRCh38
rs759977426 Chr.10: 24595047-24595048 on Build GRCh38
rs544929235 Chr.10: 24595186-24595186 on Build GRCh38
rs757386522 Chr.10: 24595054-24595055 on Build GRCh38
rs780397561 Chr.10: 24595106-24595106 on Build GRCh38
rs376880366 Chr.10: 24595101-24595101 on Build GRCh38
rs746542993 Chr.10: 24595068-24595068 on Build GRCh38
rs1001390982 Chr.10: 24595208-24595212 on Build GRCh38
rs1006357701 Chr.10: 24595143-24595143 on Build GRCh38
rs374020863 Chr.10: 24595089-24595089 on Build GRCh38
rs531270095 Chr.10: 24595103-24595103 on Build GRCh38
rs759950309 Chr.10: 24595148-24595156 on Build GRCh38
rs143518253 Chr.10: 24595102-24595102 on Build GRCh38
rs137887968 Chr.10: 24595227-24595227 on Build GRCh38
rs752386473 Chr.10: 24595061-24595061 on Build GRCh38
rs375712686 Chr.10: 24595196-24595196 on Build GRCh38
rs958991304 Chr.10: 24595152-24595152 on Build GRCh38
rs747253668 Chr.10: 24595109-24595109 on Build GRCh38
rs751043298 Chr.10: 24595053-24595053 on Build GRCh38
rs924128075 Chr.10: 24595225-24595225 on Build GRCh38
rs370081996 Chr.10: 24595224-24595224 on Build GRCh38
rs796474813 Chr.10: 24595047-24595048 on Build GRCh38
rs781504179 Chr.10: 24595099-24595100 on Build GRCh38
rs545302973 Chr.10: 24595072-24595072 on Build GRCh38
rs1046725567 Chr.10: 24595056-24595056 on Build GRCh38
rs759040226 Chr.10: 24595049-24595050 on Build GRCh38
rs757148348 Chr.10: 24595091-24595091 on Build GRCh38
rs752612284 Chr.10: 24595084-24595084 on Build GRCh38
rs375121801 Chr.10: 24595232-24595232 on Build GRCh38
rs1005078878 Chr.10: 24595069-24595069 on Build GRCh38
rs1025227407 Chr.10: 24595179-24595179 on Build GRCh38
rs777140359 Chr.10: 24595150-24595150 on Build GRCh38
rs767154942 Chr.10: 24595054-24595054 on Build GRCh38
rs537787071 Chr.10: 24595163-24595163 on Build GRCh38
rs1029459359 Chr.10: 24595181-24595181 on Build GRCh38
rs757139557 Chr.10: 24595230-24595230 on Build GRCh38
rs1011920114 Chr.10: 24595158-24595158 on Build GRCh38
rs773838375 Chr.10: 24595182-24595182 on Build GRCh38
rs751735287 Chr.10: 24595052-24595053 on Build GRCh38
rs745596228 Chr.10: 24595202-24595204 on Build GRCh38
rs767063390 Chr.10: 24595190-24595190 on Build GRCh38
rs368928428 Chr.10: 24595073-24595073 on Build GRCh38

View Full Product Details