Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 27341237-27341464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC34
Entrez Gene ID
91057
Gene Name
coiled-coil domain containing 34
Gene Aliases
L15, NY-REN-41, RAMA3
UniGene
Hs.143733
Gene Chromosome Location
Chr.11: 27338514-27363248 on Build GRCh38
Transcripts
NM_030771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5159961 c.733G>C p.A245P Chr.11: 27341424-27341424 on Build GRCh38
6309321 c.723G>A p.K241K Chr.11: 27341434-27341434 on Build GRCh38
2113301 c.765+1G>T p.? Chr.11: 27341391-27341391 on Build GRCh38
1353323 c.731delA p.N244fs*28 Chr.11: 27341426-27341426 on Build GRCh38
1353322 c.731_732insA p.N244fs*3 Chr.11: 27341425-27341426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903195133 Chr.11: 27341301-27341301 on Build GRCh38
rs757202932 Chr.11: 27341391-27341391 on Build GRCh38
rs750793174 Chr.11: 27341368-27341368 on Build GRCh38
rs899886720 Chr.11: 27341353-27341353 on Build GRCh38
rs922999405 Chr.11: 27341426-27341426 on Build GRCh38
rs368533590 Chr.11: 27341269-27341271 on Build GRCh38
rs192684924 Chr.11: 27341334-27341334 on Build GRCh38
rs200519703 Chr.11: 27341377-27341377 on Build GRCh38
rs778642826 Chr.11: 27341407-27341407 on Build GRCh38
rs753714609 Chr.11: 27341384-27341384 on Build GRCh38
rs942597919 Chr.11: 27341338-27341338 on Build GRCh38
rs556960386 Chr.11: 27341380-27341380 on Build GRCh38
rs949996651 Chr.11: 27341289-27341289 on Build GRCh38
rs771674483 Chr.11: 27341426-27341426 on Build GRCh38
rs746548730 Chr.11: 27341429-27341429 on Build GRCh38
rs771808291 Chr.11: 27341342-27341343 on Build GRCh38
rs1000159123 Chr.11: 27341332-27341332 on Build GRCh38
rs367564446 Chr.11: 27341416-27341416 on Build GRCh38
rs745647709 Chr.11: 27341422-27341422 on Build GRCh38
rs12282639 Chr.11: 27341370-27341370 on Build GRCh38
rs751660219 Chr.11: 27341371-27341371 on Build GRCh38
rs779808426 Chr.11: 27341427-27341427 on Build GRCh38
rs553293015 Chr.11: 27341343-27341343 on Build GRCh38
rs773019149 Chr.11: 27341425-27341426 on Build GRCh38
rs774898433 Chr.11: 27341282-27341282 on Build GRCh38
rs1051021693 Chr.11: 27341333-27341333 on Build GRCh38
rs924012640 Chr.11: 27341285-27341285 on Build GRCh38
rs571054649 Chr.11: 27341295-27341295 on Build GRCh38
rs202073442 Chr.11: 27341360-27341360 on Build GRCh38
rs765464387 Chr.11: 27341361-27341361 on Build GRCh38

View Full Product Details