Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 688272-688501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEAF1
Entrez Gene ID
10522
Gene Name
DEAF1, transcription factor
Gene Aliases
MRD24, NUDR, SPN, ZMYND5
UniGene
Hs.243994
Gene Chromosome Location
Chr.11: 644220-695754 on Build GRCh38
Transcripts
NM_001293634.1 NM_021008.3 XM_011519842.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4887909 c.489G>A p.L163L Chr.11: 688359-688359 on Build GRCh38
6190767 c.438G>A p.A146A Chr.11: 688410-688410 on Build GRCh38
1249926 c.396G>A p.R132R Chr.11: 688452-688452 on Build GRCh38
6642105 c.484G>A p.G162R Chr.11: 688364-688364 on Build GRCh38
4974523 c.438G>A p.A146A Chr.11: 688410-688410 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745760142 Chr.11: 688313-688313 on Build GRCh38
rs938689506 Chr.11: 688385-688385 on Build GRCh38
rs780138163 Chr.11: 688468-688468 on Build GRCh38
rs375575398 Chr.11: 688338-688338 on Build GRCh38
rs756048516 Chr.11: 688467-688467 on Build GRCh38
rs7116631 Chr.11: 688312-688312 on Build GRCh38
rs555185365 Chr.11: 688318-688318 on Build GRCh38
rs138629550 Chr.11: 688303-688303 on Build GRCh38
rs754319706 Chr.11: 688329-688329 on Build GRCh38
rs1027561012 Chr.11: 688346-688346 on Build GRCh38
rs200666406 Chr.11: 688434-688434 on Build GRCh38
rs765539686 Chr.11: 688337-688337 on Build GRCh38
rs765741994 Chr.11: 688420-688420 on Build GRCh38
rs200600001 Chr.11: 688376-688376 on Build GRCh38
rs776719665 Chr.11: 688418-688418 on Build GRCh38
rs754837453 Chr.11: 688476-688476 on Build GRCh38
rs756731135 Chr.11: 688351-688351 on Build GRCh38
rs775316641 Chr.11: 688316-688316 on Build GRCh38
rs777825063 Chr.11: 688302-688302 on Build GRCh38
rs566174877 Chr.11: 688416-688416 on Build GRCh38
rs369913665 Chr.11: 688410-688410 on Build GRCh38
rs574863473 Chr.11: 688350-688350 on Build GRCh38
rs760044921 Chr.11: 688336-688336 on Build GRCh38
rs773559333 Chr.11: 688398-688398 on Build GRCh38
rs1043593952 Chr.11: 688469-688469 on Build GRCh38
rs11552850 Chr.11: 688334-688334 on Build GRCh38
rs141368610 Chr.11: 688364-688364 on Build GRCh38
rs201490761 Chr.11: 688481-688481 on Build GRCh38
rs369913079 Chr.11: 688323-688323 on Build GRCh38
rs760007246 Chr.11: 688419-688419 on Build GRCh38
rs775897060 Chr.11: 688431-688431 on Build GRCh38
rs199729614 Chr.11: 688449-688449 on Build GRCh38
rs373131091 Chr.11: 688437-688437 on Build GRCh38
rs749155933 Chr.11: 688475-688475 on Build GRCh38
rs769481120 Chr.11: 688315-688315 on Build GRCh38
rs777396337 Chr.11: 688343-688343 on Build GRCh38
rs751217884 Chr.11: 688447-688447 on Build GRCh38
rs373778878 Chr.11: 688324-688324 on Build GRCh38
rs373316580 Chr.11: 688480-688480 on Build GRCh38
rs749062794 Chr.11: 688387-688387 on Build GRCh38
rs187623972 Chr.11: 688365-688365 on Build GRCh38
rs749927951 Chr.11: 688465-688465 on Build GRCh38
rs545790696 Chr.11: 688456-688456 on Build GRCh38
rs772491770 Chr.11: 688391-688391 on Build GRCh38

View Full Product Details