Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 37562106-37562331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNALI1
Entrez Gene ID
7802
Gene Name
dynein axonemal light intermediate chain 1
Gene Aliases
P28, dJ423B22.5, hp28
UniGene
Hs.406050
Gene Chromosome Location
Chr.1: 37556919-37566857 on Build GRCh38
Transcripts
NM_003462.3 XM_005271172.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7365125 c.757A>C p.K253Q Chr.1: 37562195-37562195 on Build GRCh38
6246128 c.747G>T p.V249V Chr.1: 37562185-37562185 on Build GRCh38
5030912 c.752_754delAGA p.K253delK Chr.1: 37562190-37562192 on Build GRCh38
6647436 c.793A>C p.N265H Chr.1: 37562231-37562231 on Build GRCh38
3400732 c.671A>G p.D224G Chr.1: 37562109-37562109 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780420246 Chr.1: 37562254-37562254 on Build GRCh38
rs74476048 Chr.1: 37562255-37562255 on Build GRCh38
rs372987118 Chr.1: 37562135-37562135 on Build GRCh38
rs770080333 Chr.1: 37562266-37562266 on Build GRCh38
rs769354887 Chr.1: 37562188-37562188 on Build GRCh38
rs200294729 Chr.1: 37562171-37562171 on Build GRCh38
rs752655458 Chr.1: 37562230-37562230 on Build GRCh38
rs764857869 Chr.1: 37562139-37562139 on Build GRCh38
rs750872461 Chr.1: 37562178-37562178 on Build GRCh38
rs763908230 Chr.1: 37562237-37562237 on Build GRCh38
rs763916437 Chr.1: 37562288-37562288 on Build GRCh38
rs750070808 Chr.1: 37562142-37562142 on Build GRCh38
rs751524648 Chr.1: 37562162-37562162 on Build GRCh38
rs199858395 Chr.1: 37562165-37562165 on Build GRCh38
rs755088083 Chr.1: 37562256-37562256 on Build GRCh38
rs757201027 Chr.1: 37562190-37562192 on Build GRCh38
rs775319419 Chr.1: 37562279-37562279 on Build GRCh38
rs200945209 Chr.1: 37562163-37562163 on Build GRCh38
rs146033675 Chr.1: 37562273-37562273 on Build GRCh38
rs766981306 Chr.1: 37562277-37562277 on Build GRCh38
rs376695154 Chr.1: 37562242-37562242 on Build GRCh38
rs779051650 Chr.1: 37562175-37562175 on Build GRCh38
rs146461161 Chr.1: 37562169-37562169 on Build GRCh38
rs369164958 Chr.1: 37562179-37562179 on Build GRCh38
rs772773309 Chr.1: 37562211-37562211 on Build GRCh38
rs547699738 Chr.1: 37562295-37562295 on Build GRCh38
rs748939264 Chr.1: 37562267-37562267 on Build GRCh38
rs373348056 Chr.1: 37562203-37562203 on Build GRCh38
rs757356271 Chr.1: 37562161-37562161 on Build GRCh38
rs773779792 Chr.1: 37562219-37562219 on Build GRCh38
rs755900637 Chr.1: 37562234-37562234 on Build GRCh38
rs762414373 Chr.1: 37562153-37562153 on Build GRCh38
rs41267313 Chr.1: 37562195-37562195 on Build GRCh38
rs142558790 Chr.1: 37562214-37562214 on Build GRCh38
rs1006411459 Chr.1: 37562221-37562221 on Build GRCh38
rs780347969 Chr.1: 37562212-37562212 on Build GRCh38
rs758626747 Chr.1: 37562246-37562246 on Build GRCh38
rs749047646 Chr.1: 37562185-37562187 on Build GRCh38
rs772091493 Chr.1: 37562287-37562288 on Build GRCh38
rs781592854 Chr.1: 37562264-37562264 on Build GRCh38
rs751802740 Chr.1: 37562291-37562291 on Build GRCh38
rs142372828 Chr.1: 37562268-37562269 on Build GRCh38
rs942711965 Chr.1: 37562304-37562304 on Build GRCh38
rs376104831 Chr.1: 37562289-37562289 on Build GRCh38
rs760611244 Chr.1: 37562281-37562281 on Build GRCh38
rs745929528 Chr.1: 37562281-37562281 on Build GRCh38
rs138550216 Chr.1: 37562136-37562136 on Build GRCh38
rs527849851 Chr.1: 37562268-37562268 on Build GRCh38
rs148524948 Chr.1: 37562177-37562177 on Build GRCh38
rs747523454 Chr.1: 37562187-37562187 on Build GRCh38
rs753734584 Chr.1: 37562287-37562287 on Build GRCh38
rs144278053 Chr.1: 37562170-37562170 on Build GRCh38
rs759038962 Chr.1: 37562225-37562225 on Build GRCh38
rs978629805 Chr.1: 37562144-37562144 on Build GRCh38
rs769290870 Chr.1: 37562300-37562300 on Build GRCh38
rs780891234 Chr.1: 37562282-37562282 on Build GRCh38
rs868438389 Chr.1: 37562172-37562172 on Build GRCh38
rs777641442 Chr.1: 37562144-37562145 on Build GRCh38
rs199870103 Chr.1: 37562226-37562226 on Build GRCh38
rs773976202 Chr.1: 37562272-37562272 on Build GRCh38
rs770166442 Chr.1: 37562206-37562206 on Build GRCh38
rs762620270 Chr.1: 37562213-37562213 on Build GRCh38
rs1004694989 Chr.1: 37562257-37562257 on Build GRCh38

View Full Product Details