Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70253736-70253995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AARS
Entrez Gene ID
16
Gene Name
alanyl-tRNA synthetase
Gene Aliases
CMT2N, EIEE29
UniGene
Hs.315137
Gene Chromosome Location
Chr.16: 70252295-70289552 on Build GRCh38
Transcripts
NM_001605.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3672316 c.2447G>T p.R816L Chr.16: 70253992-70253992 on Build GRCh38
4644011 c.2505C>T p.A835A Chr.16: 70253934-70253934 on Build GRCh38
6589288 c.2521-8delT p.? Chr.16: 70253808-70253808 on Build GRCh38
972985 c.2548G>A p.D850N Chr.16: 70253773-70253773 on Build GRCh38
4434968 c.2527G>A p.E843K Chr.16: 70253794-70253794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs575239120 Chr.16: 70253771-70253771 on Build GRCh38
rs756241399 Chr.16: 70253834-70253834 on Build GRCh38
rs750566424 Chr.16: 70253828-70253828 on Build GRCh38
rs1007113248 Chr.16: 70253911-70253911 on Build GRCh38
rs368011194 Chr.16: 70253769-70253769 on Build GRCh38
rs767463251 Chr.16: 70253822-70253822 on Build GRCh38
rs759350421 Chr.16: 70253890-70253891 on Build GRCh38
rs942383684 Chr.16: 70253819-70253819 on Build GRCh38
rs1018549676 Chr.16: 70253917-70253917 on Build GRCh38
rs111333360 Chr.16: 70253802-70253802 on Build GRCh38
rs200586605 Chr.16: 70253803-70253803 on Build GRCh38
rs772849771 Chr.16: 70253773-70253773 on Build GRCh38
rs770634653 Chr.16: 70253786-70253786 on Build GRCh38
rs766300063 Chr.16: 70253846-70253847 on Build GRCh38
rs199521984 Chr.16: 70253810-70253810 on Build GRCh38
rs766741526 Chr.16: 70253842-70253842 on Build GRCh38
rs754189661 Chr.16: 70253843-70253843 on Build GRCh38
rs111980438 Chr.16: 70253801-70253801 on Build GRCh38
rs777095182 Chr.16: 70253885-70253885 on Build GRCh38
rs891381242 Chr.16: 70253910-70253910 on Build GRCh38
rs962230274 Chr.16: 70253768-70253768 on Build GRCh38
rs897597773 Chr.16: 70253863-70253863 on Build GRCh38
rs745577157 Chr.16: 70253931-70253931 on Build GRCh38
rs751391874 Chr.16: 70253847-70253847 on Build GRCh38
rs762777733 Chr.16: 70253947-70253947 on Build GRCh38
rs760816488 Chr.16: 70253954-70253954 on Build GRCh38
rs754119012 Chr.16: 70253971-70253971 on Build GRCh38
rs746615745 Chr.16: 70253774-70253774 on Build GRCh38
rs371962301 Chr.16: 70253850-70253850 on Build GRCh38
rs371712914 Chr.16: 70253789-70253789 on Build GRCh38
rs541254641 Chr.16: 70253856-70253856 on Build GRCh38
rs746557071 Chr.16: 70253899-70253899 on Build GRCh38
rs1041532704 Chr.16: 70253895-70253895 on Build GRCh38
rs928532751 Chr.16: 70253777-70253777 on Build GRCh38
rs767164973 Chr.16: 70253957-70253958 on Build GRCh38
rs941883610 Chr.16: 70253825-70253825 on Build GRCh38
rs200328881 Chr.16: 70253812-70253812 on Build GRCh38
rs775304067 Chr.16: 70253936-70253936 on Build GRCh38
rs192447854 Chr.16: 70253851-70253851 on Build GRCh38
rs758988205 Chr.16: 70253852-70253852 on Build GRCh38
rs577885582 Chr.16: 70253785-70253785 on Build GRCh38
rs766651955 Chr.16: 70253963-70253963 on Build GRCh38
rs769571173 Chr.16: 70253934-70253934 on Build GRCh38
rs540218496 Chr.16: 70253865-70253865 on Build GRCh38
rs376619331 Chr.16: 70253900-70253900 on Build GRCh38
rs762934931 Chr.16: 70253797-70253802 on Build GRCh38
rs776276100 Chr.16: 70253920-70253920 on Build GRCh38

View Full Product Details