Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67150441-67150686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT9
Entrez Gene ID
84752
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 9
Gene Aliases
-
UniGene
Hs.744158
Gene Chromosome Location
Chr.16: 67148102-67150999 on Build GRCh38
Transcripts
NM_033309.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3691094 c.24C>A p.R8R Chr.16: 67150462-67150462 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs955154023 Chr.16: 67150640-67150640 on Build GRCh38
rs962243718 Chr.16: 67150625-67150625 on Build GRCh38
rs994249840 Chr.16: 67150632-67150634 on Build GRCh38
rs1045738981 Chr.16: 67150536-67150536 on Build GRCh38
rs887984046 Chr.16: 67150610-67150610 on Build GRCh38
rs761378828 Chr.16: 67150489-67150489 on Build GRCh38
rs1029247785 Chr.16: 67150635-67150635 on Build GRCh38
rs911007005 Chr.16: 67150460-67150460 on Build GRCh38
rs546732775 Chr.16: 67150634-67150634 on Build GRCh38
rs762446480 Chr.16: 67150474-67150476 on Build GRCh38
rs952592388 Chr.16: 67150649-67150649 on Build GRCh38
rs1006383377 Chr.16: 67150614-67150614 on Build GRCh38
rs1043112388 Chr.16: 67150607-67150607 on Build GRCh38
rs890695799 Chr.16: 67150619-67150619 on Build GRCh38
rs926043429 Chr.16: 67150612-67150612 on Build GRCh38
rs1001991444 Chr.16: 67150578-67150578 on Build GRCh38
rs890726829 Chr.16: 67150474-67150474 on Build GRCh38
rs749983826 Chr.16: 67150499-67150499 on Build GRCh38
rs775502891 Chr.16: 67150464-67150464 on Build GRCh38
rs1009138502 Chr.16: 67150622-67150622 on Build GRCh38
rs959972058 Chr.16: 67150581-67150581 on Build GRCh38
rs764117502 Chr.16: 67150477-67150477 on Build GRCh38
rs767311241 Chr.16: 67150498-67150498 on Build GRCh38
rs769419679 Chr.16: 67150462-67150462 on Build GRCh38
rs932147253 Chr.16: 67150616-67150616 on Build GRCh38
rs1034529487 Chr.16: 67150591-67150591 on Build GRCh38
rs985652911 Chr.16: 67150666-67150666 on Build GRCh38
rs896788573 Chr.16: 67150629-67150629 on Build GRCh38
rs1019268863 Chr.16: 67150496-67150496 on Build GRCh38
rs946032858 Chr.16: 67150590-67150590 on Build GRCh38
rs935845412 Chr.16: 67150466-67150466 on Build GRCh38
rs763052097 Chr.16: 67150471-67150471 on Build GRCh38
rs961104381 Chr.16: 67150497-67150497 on Build GRCh38
rs907298659 Chr.16: 67150572-67150572 on Build GRCh38
rs868614729 Chr.16: 67150470-67150470 on Build GRCh38
rs992819222 Chr.16: 67150588-67150589 on Build GRCh38
rs534741183 Chr.16: 67150617-67150617 on Build GRCh38
rs913274876 Chr.16: 67150589-67150589 on Build GRCh38
rs980357217 Chr.16: 67150567-67150567 on Build GRCh38
rs1007831742 Chr.16: 67150476-67150476 on Build GRCh38
rs774057583 Chr.16: 67150481-67150481 on Build GRCh38
rs952545269 Chr.16: 67150545-67150545 on Build GRCh38
rs761101988 Chr.16: 67150514-67150514 on Build GRCh38
rs372144037 Chr.16: 67150529-67150529 on Build GRCh38

View Full Product Details