Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 93710014-93710239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM133A
Entrez Gene ID
286499
Gene Name
family with sequence similarity 133 member A
Gene Aliases
CT115
UniGene
Hs.110069
Gene Chromosome Location
Chr.X: 93673730-93712274 on Build GRCh38
Transcripts
NM_001171109.1 NM_001171110.1 NM_001171111.1 NM_173698.2 XM_006724641.3 XM_011530925.2 XM_011530926.2 XM_011530927.2 XM_011530928.1 XM_011530929.1 XM_011530931.2 XM_017029454.1 XM_017029455.1 XM_017029456.1

SNP Detail

SNP ID Chromosome Location
rs749386717 Chr.X: 93710146-93710146 on Build GRCh38
rs201513170 Chr.X: 93710065-93710065 on Build GRCh38
rs768851012 Chr.X: 93710155-93710155 on Build GRCh38
rs760979272 Chr.X: 93710199-93710199 on Build GRCh38
rs1014787716 Chr.X: 93710203-93710203 on Build GRCh38
rs745615374 Chr.X: 93710107-93710107 on Build GRCh38
rs778405212 Chr.X: 93710118-93710118 on Build GRCh38
rs753406171 Chr.X: 93710096-93710096 on Build GRCh38
rs200658365 Chr.X: 93710197-93710197 on Build GRCh38
rs895822158 Chr.X: 93710047-93710049 on Build GRCh38
rs780369284 Chr.X: 93710142-93710142 on Build GRCh38
rs753503341 Chr.X: 93710086-93710088 on Build GRCh38
rs754447873 Chr.X: 93710097-93710097 on Build GRCh38
rs778847468 Chr.X: 93710176-93710176 on Build GRCh38
rs762136693 Chr.X: 93710081-93710081 on Build GRCh38
rs745706648 Chr.X: 93710119-93710122 on Build GRCh38
rs756097094 Chr.X: 93710202-93710203 on Build GRCh38
rs887737389 Chr.X: 93710187-93710187 on Build GRCh38
rs999991546 Chr.X: 93710165-93710165 on Build GRCh38
rs766045732 Chr.X: 93710085-93710086 on Build GRCh38
rs757117737 Chr.X: 93710084-93710084 on Build GRCh38
rs751570752 Chr.X: 93710082-93710082 on Build GRCh38
rs748740241 Chr.X: 93710191-93710191 on Build GRCh38
rs756864294 Chr.X: 93710086-93710091 on Build GRCh38
rs756422345 Chr.X: 93710141-93710141 on Build GRCh38
rs752226811 Chr.X: 93710061-93710061 on Build GRCh38
rs201987125 Chr.X: 93710198-93710198 on Build GRCh38
rs911541951 Chr.X: 93710206-93710207 on Build GRCh38

View Full Product Details