Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6498808-6499043 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNHD1
Entrez Gene ID
144132
Gene Name
dynein heavy chain domain 1
Gene Aliases
C11orf47, CCDC35, DHCD1, DNHD1L
UniGene
Hs.720080
Gene Chromosome Location
Chr.11: 6497296-6572025 on Build GRCh38
Transcripts
NM_144666.2 NM_173589.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4035397 c.722A>T p.E241V Chr.11: 6498937-6498937 on Build GRCh38
5051017 c.723G>A p.E241E Chr.11: 6498938-6498938 on Build GRCh38
4418283 c.719T>G p.V240G Chr.11: 6498934-6498934 on Build GRCh38
348514 c.651C>G p.L217L Chr.11: 6498866-6498866 on Build GRCh38
1746463 c.739G>C p.E247Q Chr.11: 6498954-6498954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201927350 Chr.11: 6498899-6498899 on Build GRCh38
rs375885854 Chr.11: 6498989-6498989 on Build GRCh38
rs766184497 Chr.11: 6498980-6498980 on Build GRCh38
rs2555158 Chr.11: 6498934-6498934 on Build GRCh38
rs1027453410 Chr.11: 6498854-6498854 on Build GRCh38
rs756233062 Chr.11: 6498996-6498996 on Build GRCh38
rs749817858 Chr.11: 6498916-6498916 on Build GRCh38
rs767570130 Chr.11: 6498990-6498990 on Build GRCh38
rs762966208 Chr.11: 6498973-6498973 on Build GRCh38
rs777933250 Chr.11: 6498910-6498910 on Build GRCh38
rs774239043 Chr.11: 6498984-6498984 on Build GRCh38
rs762349463 Chr.11: 6498873-6498873 on Build GRCh38
rs767766408 Chr.11: 6498904-6498904 on Build GRCh38
rs764543070 Chr.11: 6499002-6499002 on Build GRCh38
rs35423081 Chr.11: 6498834-6498834 on Build GRCh38
rs780027381 Chr.11: 6498925-6498925 on Build GRCh38
rs759660506 Chr.11: 6498987-6498987 on Build GRCh38
rs140861406 Chr.11: 6498829-6498829 on Build GRCh38
rs757658550 Chr.11: 6498930-6498930 on Build GRCh38
rs773264422 Chr.11: 6498887-6498887 on Build GRCh38
rs752893246 Chr.11: 6498909-6498909 on Build GRCh38
rs922151274 Chr.11: 6498971-6498971 on Build GRCh38
rs776488073 Chr.11: 6498935-6498935 on Build GRCh38
rs768386552 Chr.11: 6498835-6498835 on Build GRCh38
rs201431169 Chr.11: 6498995-6498995 on Build GRCh38
rs200724241 Chr.11: 6498892-6498892 on Build GRCh38
rs752921716 Chr.11: 6498994-6498994 on Build GRCh38
rs751588566 Chr.11: 6498900-6498900 on Build GRCh38
rs746560822 Chr.11: 6498933-6498933 on Build GRCh38
rs769631973 Chr.11: 6498962-6498962 on Build GRCh38
rs780934118 Chr.11: 6498842-6498842 on Build GRCh38
rs142166265 Chr.11: 6498901-6498901 on Build GRCh38
rs767420341 Chr.11: 6498886-6498886 on Build GRCh38
rs761752956 Chr.11: 6498876-6498876 on Build GRCh38
rs888932802 Chr.11: 6498964-6498964 on Build GRCh38
rs554331237 Chr.11: 6498942-6498942 on Build GRCh38

View Full Product Details