Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 13623736-13624009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGFL6
Entrez Gene ID
25975
Gene Name
EGF like domain multiple 6
Gene Aliases
MAEG, W80
UniGene
Hs.12844
Gene Chromosome Location
Chr.X: 13569575-13633575 on Build GRCh38
Transcripts
NM_001167890.1 NM_015507.3

SNP Detail

SNP ID Chromosome Location
rs372552929 Chr.X: 13623882-13623882 on Build GRCh38
rs768721759 Chr.X: 13623959-13623959 on Build GRCh38
rs767962439 Chr.X: 13623916-13623916 on Build GRCh38
rs753342813 Chr.X: 13623957-13623957 on Build GRCh38
rs142725131 Chr.X: 13623852-13623852 on Build GRCh38
rs990302211 Chr.X: 13623791-13623791 on Build GRCh38
rs147554528 Chr.X: 13623969-13623969 on Build GRCh38
rs772077108 Chr.X: 13623972-13623972 on Build GRCh38
rs1021823794 Chr.X: 13623830-13623830 on Build GRCh38
rs780017714 Chr.X: 13623837-13623837 on Build GRCh38
rs201626449 Chr.X: 13623931-13623931 on Build GRCh38
rs977738246 Chr.X: 13623844-13623844 on Build GRCh38
rs774648370 Chr.X: 13623892-13623892 on Build GRCh38
rs745670028 Chr.X: 13623802-13623802 on Build GRCh38
rs866466116 Chr.X: 13623880-13623880 on Build GRCh38
rs769889467 Chr.X: 13623856-13623856 on Build GRCh38
rs375448294 Chr.X: 13623917-13623917 on Build GRCh38
rs921319364 Chr.X: 13623934-13623934 on Build GRCh38
rs897331582 Chr.X: 13623809-13623809 on Build GRCh38
rs762208309 Chr.X: 13623977-13623977 on Build GRCh38
rs759918341 Chr.X: 13623910-13623910 on Build GRCh38
rs1033410455 Chr.X: 13623761-13623761 on Build GRCh38
rs771944061 Chr.X: 13623804-13623804 on Build GRCh38
rs967622558 Chr.X: 13623836-13623836 on Build GRCh38
rs368591398 Chr.X: 13623824-13623824 on Build GRCh38
rs773206930 Chr.X: 13623857-13623857 on Build GRCh38
rs756869065 Chr.X: 13623794-13623794 on Build GRCh38
rs957785835 Chr.X: 13623774-13623774 on Build GRCh38
rs775690513 Chr.X: 13623812-13623812 on Build GRCh38
rs749522199 Chr.X: 13623876-13623876 on Build GRCh38
rs376844477 Chr.X: 13623784-13623784 on Build GRCh38
rs761780042 Chr.X: 13623809-13623809 on Build GRCh38
rs761225255 Chr.X: 13623925-13623925 on Build GRCh38

View Full Product Details