Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31139669-31139934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSC1
Entrez Gene ID
1823
Gene Name
desmocollin 1
Gene Aliases
CDHF1, DG2/DG3
UniGene
Hs.567260
Gene Chromosome Location
Chr.18: 31129236-31162856 on Build GRCh38
Transcripts
NM_004948.3 NM_024421.2 NR_110785.1

Gene Symbol
DSCAS
Entrez Gene ID
101927698
Gene Name
DSC1/DSC2 antisense RNA
Gene Aliases
-
UniGene
Hs.579538
Gene Chromosome Location
Chr.18: 31101588-31162789 on Build GRCh38
Transcripts
NM_004948.3 NM_024421.2 NR_110785.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
987491 c.1606G>T p.E536* Chr.18: 31139805-31139805 on Build GRCh38
1711102 c.1615T>C p.F539L Chr.18: 31139796-31139796 on Build GRCh38
5106447 c.1625_1626insA p.N542fs*28 Chr.18: 31139785-31139786 on Build GRCh38
987490 c.1606G>T p.E536* Chr.18: 31139805-31139805 on Build GRCh38
5106446 c.1625_1626insA p.N542fs*28 Chr.18: 31139785-31139786 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140152065 Chr.18: 31139857-31139857 on Build GRCh38
rs772930554 Chr.18: 31139834-31139834 on Build GRCh38
rs750569072 Chr.18: 31139906-31139906 on Build GRCh38
rs763321997 Chr.18: 31139868-31139868 on Build GRCh38
rs774348552 Chr.18: 31139704-31139704 on Build GRCh38
rs758244714 Chr.18: 31139774-31139774 on Build GRCh38
rs753101721 Chr.18: 31139742-31139742 on Build GRCh38
rs749661572 Chr.18: 31139782-31139784 on Build GRCh38
rs770148538 Chr.18: 31139700-31139700 on Build GRCh38
rs765718498 Chr.18: 31139734-31139734 on Build GRCh38
rs750956702 Chr.18: 31139888-31139888 on Build GRCh38
rs746574734 Chr.18: 31139836-31139836 on Build GRCh38
rs761675425 Chr.18: 31139705-31139705 on Build GRCh38
rs769195695 Chr.18: 31139817-31139817 on Build GRCh38
rs757482342 Chr.18: 31139881-31139881 on Build GRCh38
rs111561499 Chr.18: 31139821-31139821 on Build GRCh38
rs764323087 Chr.18: 31139756-31139756 on Build GRCh38
rs747983579 Chr.18: 31139813-31139813 on Build GRCh38
rs781198190 Chr.18: 31139779-31139779 on Build GRCh38
rs758723937 Chr.18: 31139752-31139752 on Build GRCh38
rs770611900 Chr.18: 31139842-31139842 on Build GRCh38
rs757366419 Chr.18: 31139775-31139775 on Build GRCh38
rs200980589 Chr.18: 31139904-31139904 on Build GRCh38
rs911117236 Chr.18: 31139741-31139741 on Build GRCh38
rs777747761 Chr.18: 31139902-31139902 on Build GRCh38
rs755012258 Chr.18: 31139797-31139797 on Build GRCh38
rs775927656 Chr.18: 31139864-31139864 on Build GRCh38
rs745950187 Chr.18: 31139784-31139784 on Build GRCh38
rs772697260 Chr.18: 31139707-31139707 on Build GRCh38
rs751952855 Chr.18: 31139876-31139876 on Build GRCh38
rs113931359 Chr.18: 31139863-31139863 on Build GRCh38
rs766317004 Chr.18: 31139828-31139828 on Build GRCh38
rs760101651 Chr.18: 31139720-31139720 on Build GRCh38
rs534535281 Chr.18: 31139726-31139726 on Build GRCh38
rs1789043 Chr.18: 31139711-31139711 on Build GRCh38
rs768713268 Chr.18: 31139701-31139701 on Build GRCh38
rs946546168 Chr.18: 31139760-31139760 on Build GRCh38
rs143317791 Chr.18: 31139822-31139822 on Build GRCh38
rs115338830 Chr.18: 31139849-31139849 on Build GRCh38
rs778960561 Chr.18: 31139809-31139809 on Build GRCh38
rs751679267 Chr.18: 31139770-31139770 on Build GRCh38
rs771888120 Chr.18: 31139706-31139706 on Build GRCh38
rs765748865 Chr.18: 31139710-31139710 on Build GRCh38
rs1043568088 Chr.18: 31139826-31139826 on Build GRCh38
rs372768585 Chr.18: 31139835-31139835 on Build GRCh38
rs775924706 Chr.18: 31139781-31139781 on Build GRCh38
rs760251396 Chr.18: 31139708-31139708 on Build GRCh38
rs774887084 Chr.18: 31139902-31139920 on Build GRCh38

View Full Product Details