Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 158255837-158256102 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD1A
Entrez Gene ID
909
Gene Name
CD1a molecule
Gene Aliases
CD1, FCB6, HTA1, R4, T6
UniGene
Hs.1309
Gene Chromosome Location
Chr.1: 158248329-158258269 on Build GRCh38
Transcripts
NM_001320652.1 NM_001763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
358228 c.355T>C p.C119R Chr.1: 158256033-158256033 on Build GRCh38
1157858 c.371G>T p.G124V Chr.1: 158256049-158256049 on Build GRCh38
6121587 c.326-1G>C p.? Chr.1: 158256003-158256003 on Build GRCh38
83937 c.328C>T p.P110S Chr.1: 158256006-158256006 on Build GRCh38
676213 c.362T>G p.L121R Chr.1: 158256040-158256040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930221944 Chr.1: 158256034-158256034 on Build GRCh38
rs112853675 Chr.1: 158256069-158256069 on Build GRCh38
rs769926346 Chr.1: 158255958-158255958 on Build GRCh38
rs773208720 Chr.1: 158255962-158255962 on Build GRCh38
rs372236626 Chr.1: 158256018-158256018 on Build GRCh38
rs761067402 Chr.1: 158255967-158255967 on Build GRCh38
rs937430092 Chr.1: 158255948-158255948 on Build GRCh38
rs369938958 Chr.1: 158255965-158255965 on Build GRCh38
rs755789793 Chr.1: 158256009-158256009 on Build GRCh38
rs373036607 Chr.1: 158256010-158256010 on Build GRCh38
rs1043222177 Chr.1: 158255880-158255880 on Build GRCh38
rs766819985 Chr.1: 158255981-158255981 on Build GRCh38
rs757425882 Chr.1: 158255882-158255882 on Build GRCh38
rs954614261 Chr.1: 158255867-158255867 on Build GRCh38
rs373749635 Chr.1: 158255908-158255908 on Build GRCh38
rs927422388 Chr.1: 158255933-158255933 on Build GRCh38
rs200643675 Chr.1: 158255994-158255994 on Build GRCh38
rs758184194 Chr.1: 158256050-158256050 on Build GRCh38
rs529984822 Chr.1: 158256072-158256072 on Build GRCh38
rs769944278 Chr.1: 158256058-158256058 on Build GRCh38
rs117141061 Chr.1: 158255999-158255999 on Build GRCh38
rs777850226 Chr.1: 158256065-158256065 on Build GRCh38
rs947230744 Chr.1: 158255907-158255907 on Build GRCh38
rs142041113 Chr.1: 158255892-158255892 on Build GRCh38
rs989148049 Chr.1: 158255869-158255869 on Build GRCh38
rs752354521 Chr.1: 158256006-158256006 on Build GRCh38
rs756288511 Chr.1: 158256060-158256060 on Build GRCh38
rs576860595 Chr.1: 158255909-158255909 on Build GRCh38
rs915999695 Chr.1: 158255860-158255860 on Build GRCh38
rs778954696 Chr.1: 158256039-158256039 on Build GRCh38
rs978801303 Chr.1: 158255921-158255921 on Build GRCh38
rs781121613 Chr.1: 158255998-158255998 on Build GRCh38
rs564719076 Chr.1: 158255953-158255953 on Build GRCh38
rs753342790 Chr.1: 158256017-158256017 on Build GRCh38
rs753262992 Chr.1: 158255966-158255967 on Build GRCh38
rs748322040 Chr.1: 158256054-158256054 on Build GRCh38
rs759854174 Chr.1: 158255997-158255997 on Build GRCh38
rs750344740 Chr.1: 158256041-158256041 on Build GRCh38
rs779894806 Chr.1: 158256053-158256053 on Build GRCh38

View Full Product Details