Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 25469789-25470036 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDCA2
Entrez Gene ID
157313
Gene Name
cell division cycle associated 2
Gene Aliases
PPP1R81, Repo-Man
UniGene
Hs.33366
Gene Chromosome Location
Chr.8: 25458569-25511308 on Build GRCh38
Transcripts
NM_001317906.1 NM_001317907.1 NM_152562.3 XM_006716294.2 XM_011544415.1 XM_011544416.2 XM_011544417.2 XM_011544418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6894796 c.736-1G>T p.? Chr.8: 25469895-25469895 on Build GRCh38
4952866 c.740C>T p.S247L Chr.8: 25469900-25469900 on Build GRCh38
6472499 c.790C>T p.P264S Chr.8: 25469950-25469950 on Build GRCh38
1200392 c.754T>C p.S252P Chr.8: 25469914-25469914 on Build GRCh38
486333 c.736A>G p.I246V Chr.8: 25469896-25469896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024110257 Chr.8: 25469816-25469816 on Build GRCh38
rs904669969 Chr.8: 25469957-25469957 on Build GRCh38
rs920438885 Chr.8: 25469822-25469822 on Build GRCh38
rs764170658 Chr.8: 25469941-25469941 on Build GRCh38
rs754321913 Chr.8: 25469944-25469944 on Build GRCh38
rs377071707 Chr.8: 25469971-25469971 on Build GRCh38
rs759882033 Chr.8: 25469936-25469936 on Build GRCh38
rs749290315 Chr.8: 25469989-25469989 on Build GRCh38
rs149146216 Chr.8: 25469905-25469905 on Build GRCh38
rs372257172 Chr.8: 25469923-25469923 on Build GRCh38
rs780717343 Chr.8: 25469962-25469962 on Build GRCh38
rs757737568 Chr.8: 25469947-25469947 on Build GRCh38
rs922533253 Chr.8: 25469932-25469932 on Build GRCh38
rs776267395 Chr.8: 25469872-25469872 on Build GRCh38
rs770965988 Chr.8: 25470000-25470000 on Build GRCh38
rs191970295 Chr.8: 25469883-25469883 on Build GRCh38
rs756163402 Chr.8: 25469940-25469940 on Build GRCh38
rs574458102 Chr.8: 25469860-25469860 on Build GRCh38
rs376009989 Chr.8: 25469917-25469917 on Build GRCh38
rs539271312 Chr.8: 25469843-25469845 on Build GRCh38
rs779198779 Chr.8: 25469852-25469852 on Build GRCh38
rs553809791 Chr.8: 25469898-25469898 on Build GRCh38
rs768265765 Chr.8: 25469866-25469866 on Build GRCh38
rs929184400 Chr.8: 25469840-25469840 on Build GRCh38
rs757647642 Chr.8: 25469850-25469850 on Build GRCh38
rs781030066 Chr.8: 25469845-25469845 on Build GRCh38
rs184433724 Chr.8: 25469953-25469953 on Build GRCh38
rs115799637 Chr.8: 25469970-25469970 on Build GRCh38
rs985320803 Chr.8: 25469843-25469843 on Build GRCh38
rs752880487 Chr.8: 25469939-25469939 on Build GRCh38
rs773025919 Chr.8: 25469899-25469899 on Build GRCh38
rs960288065 Chr.8: 25469978-25469978 on Build GRCh38
rs771539879 Chr.8: 25469877-25469878 on Build GRCh38
rs370458379 Chr.8: 25469846-25469846 on Build GRCh38
rs890418298 Chr.8: 25469881-25469881 on Build GRCh38
rs747766762 Chr.8: 25469966-25469966 on Build GRCh38
rs952146588 Chr.8: 25469824-25469824 on Build GRCh38
rs754679065 Chr.8: 25469955-25469955 on Build GRCh38
rs774268934 Chr.8: 25469921-25469921 on Build GRCh38
rs944341772 Chr.8: 25469912-25469912 on Build GRCh38

View Full Product Details