Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46809582-46809833 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf87
Entrez Gene ID
388272
Gene Name
chromosome 16 open reading frame 87
Gene Aliases
-
UniGene
Hs.406551
Gene Chromosome Location
Chr.16: 46802047-46831162 on Build GRCh38
Transcripts
NM_001001436.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4187080 c.316G>A p.A106T Chr.16: 46809633-46809633 on Build GRCh38
970790 c.291A>G p.R97R Chr.16: 46809658-46809658 on Build GRCh38
3370441 c.210A>G p.R70R Chr.16: 46809739-46809739 on Build GRCh38
3420998 c.339A>C p.E113D Chr.16: 46809610-46809610 on Build GRCh38
4770841 c.320C>T p.S107F Chr.16: 46809629-46809629 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958229315 Chr.16: 46809717-46809717 on Build GRCh38
rs752095184 Chr.16: 46809759-46809759 on Build GRCh38
rs751824128 Chr.16: 46809668-46809668 on Build GRCh38
rs370493783 Chr.16: 46809617-46809617 on Build GRCh38
rs746659744 Chr.16: 46809776-46809776 on Build GRCh38
rs759453983 Chr.16: 46809619-46809619 on Build GRCh38
rs762204685 Chr.16: 46809676-46809676 on Build GRCh38
rs780945065 Chr.16: 46809800-46809800 on Build GRCh38
rs763124975 Chr.16: 46809653-46809653 on Build GRCh38
rs550179313 Chr.16: 46809690-46809690 on Build GRCh38
rs756655129 Chr.16: 46809726-46809726 on Build GRCh38
rs777223832 Chr.16: 46809769-46809769 on Build GRCh38
rs371845990 Chr.16: 46809625-46809625 on Build GRCh38
rs750863700 Chr.16: 46809683-46809683 on Build GRCh38
rs780639684 Chr.16: 46809755-46809755 on Build GRCh38
rs767992202 Chr.16: 46809680-46809680 on Build GRCh38
rs901742965 Chr.16: 46809698-46809698 on Build GRCh38
rs767373972 Chr.16: 46809750-46809750 on Build GRCh38
rs764177597 Chr.16: 46809656-46809656 on Build GRCh38
rs770625599 Chr.16: 46809793-46809793 on Build GRCh38
rs1040674617 Chr.16: 46809662-46809662 on Build GRCh38
rs775403023 Chr.16: 46809631-46809631 on Build GRCh38

View Full Product Details