Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136722902-136723155 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM69B
Entrez Gene ID
138311
Gene Name
family with sequence similarity 69 member B
Gene Aliases
C9orf136, pp6977
UniGene
Hs.741305
Gene Chromosome Location
Chr.9: 136712572-136724718 on Build GRCh38
Transcripts
NM_152421.3 XM_006716954.3 XM_011518238.2 XM_017014273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6183147 c.537C>G p.L179L Chr.9: 136723015-136723015 on Build GRCh38
379496 c.607C>T p.R203C Chr.9: 136723085-136723085 on Build GRCh38
6229591 c.543T>C p.A181A Chr.9: 136723021-136723021 on Build GRCh38
6660073 c.625C>T p.L209L Chr.9: 136723103-136723103 on Build GRCh38
6292225 c.604C>T p.Q202* Chr.9: 136723082-136723082 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774596224 Chr.9: 136723085-136723085 on Build GRCh38
rs777710868 Chr.9: 136723121-136723121 on Build GRCh38
rs200684972 Chr.9: 136723031-136723031 on Build GRCh38
rs774543358 Chr.9: 136723131-136723131 on Build GRCh38
rs146826809 Chr.9: 136722977-136722977 on Build GRCh38
rs768332470 Chr.9: 136722974-136722974 on Build GRCh38
rs376847297 Chr.9: 136723016-136723016 on Build GRCh38
rs764306401 Chr.9: 136722994-136722994 on Build GRCh38
rs138892122 Chr.9: 136723118-136723118 on Build GRCh38
rs759786323 Chr.9: 136722925-136722925 on Build GRCh38
rs764581963 Chr.9: 136722954-136722954 on Build GRCh38
rs746860807 Chr.9: 136723124-136723124 on Build GRCh38
rs751993232 Chr.9: 136722955-136722955 on Build GRCh38
rs768951457 Chr.9: 136723084-136723084 on Build GRCh38
rs142334832 Chr.9: 136722976-136722976 on Build GRCh38
rs761251002 Chr.9: 136723094-136723094 on Build GRCh38
rs754548883 Chr.9: 136723112-136723112 on Build GRCh38
rs537431258 Chr.9: 136722979-136722979 on Build GRCh38
rs150233867 Chr.9: 136723067-136723067 on Build GRCh38
rs141104132 Chr.9: 136723041-136723041 on Build GRCh38
rs772037078 Chr.9: 136723053-136723053 on Build GRCh38
rs145651315 Chr.9: 136722991-136722991 on Build GRCh38
rs747991681 Chr.9: 136723051-136723051 on Build GRCh38
rs773704262 Chr.9: 136723093-136723093 on Build GRCh38
rs769694166 Chr.9: 136723066-136723066 on Build GRCh38
rs779684247 Chr.9: 136723030-136723030 on Build GRCh38
rs759660841 Chr.9: 136722984-136722984 on Build GRCh38
rs766502843 Chr.9: 136722921-136722921 on Build GRCh38
rs751942071 Chr.9: 136722997-136722997 on Build GRCh38
rs776979871 Chr.9: 136723041-136723041 on Build GRCh38
rs767928263 Chr.9: 136723087-136723087 on Build GRCh38
rs565704797 Chr.9: 136722963-136722963 on Build GRCh38
rs1038055511 Chr.9: 136723091-136723091 on Build GRCh38
rs765749116 Chr.9: 136723099-136723099 on Build GRCh38
rs368865388 Chr.9: 136723086-136723086 on Build GRCh38
rs747006264 Chr.9: 136723059-136723059 on Build GRCh38
rs771981811 Chr.9: 136722938-136722941 on Build GRCh38
rs1013889216 Chr.9: 136723031-136723042 on Build GRCh38
rs370469234 Chr.9: 136722992-136722992 on Build GRCh38
rs763212166 Chr.9: 136722941-136722941 on Build GRCh38
rs746012649 Chr.9: 136723097-136723099 on Build GRCh38
rs776815125 Chr.9: 136722982-136722982 on Build GRCh38
rs776868555 Chr.9: 136722922-136722922 on Build GRCh38
rs778526405 Chr.9: 136723042-136723042 on Build GRCh38
rs79974881 Chr.9: 136723018-136723018 on Build GRCh38
rs140488435 Chr.9: 136722990-136722990 on Build GRCh38
rs753009251 Chr.9: 136722928-136722928 on Build GRCh38
rs777304990 Chr.9: 136723120-136723120 on Build GRCh38
rs775386866 Chr.9: 136723135-136723136 on Build GRCh38
rs573316726 Chr.9: 136723130-136723130 on Build GRCh38
rs755566325 Chr.9: 136723029-136723029 on Build GRCh38
rs753413829 Chr.9: 136723111-136723111 on Build GRCh38
rs747935587 Chr.9: 136723116-136723118 on Build GRCh38
rs200267415 Chr.9: 136722956-136722956 on Build GRCh38
rs941082974 Chr.9: 136723090-136723090 on Build GRCh38
rs200695253 Chr.9: 136723054-136723054 on Build GRCh38
rs779956518 Chr.9: 136723128-136723128 on Build GRCh38
rs1005224702 Chr.9: 136722940-136722940 on Build GRCh38
rs11793962 Chr.9: 136722927-136722927 on Build GRCh38
rs549086212 Chr.9: 136722957-136722957 on Build GRCh38
rs534829491 Chr.9: 136722967-136722967 on Build GRCh38
rs773233628 Chr.9: 136722925-136722926 on Build GRCh38
rs149412551 Chr.9: 136723129-136723129 on Build GRCh38
rs148889146 Chr.9: 136722964-136722964 on Build GRCh38
rs553447644 Chr.9: 136723126-136723126 on Build GRCh38
rs927881394 Chr.9: 136723040-136723040 on Build GRCh38
rs754730846 Chr.9: 136722932-136722932 on Build GRCh38
rs763034680 Chr.9: 136723070-136723070 on Build GRCh38
rs752450120 Chr.9: 136723119-136723119 on Build GRCh38
rs896853300 Chr.9: 136723096-136723096 on Build GRCh38

View Full Product Details