Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 67183423-67183682 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD180
Entrez Gene ID
4064
Gene Name
CD180 molecule
Gene Aliases
LY64, Ly78, RP105
UniGene
Hs.87205
Gene Chromosome Location
Chr.5: 67182275-67196789 on Build GRCh38
Transcripts
NM_005582.2 XM_005248504.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5709742 c.1241A>G p.Q414R Chr.5: 67183602-67183602 on Build GRCh38
461961 c.1187C>T p.S396F Chr.5: 67183656-67183656 on Build GRCh38
738344 c.1342C>T p.Q448* Chr.5: 67183501-67183501 on Build GRCh38
6529528 c.1176C>G p.L392L Chr.5: 67183667-67183667 on Build GRCh38
3828327 c.1390C>T p.L464F Chr.5: 67183453-67183453 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764543994 Chr.5: 67183528-67183528 on Build GRCh38
rs769368678 Chr.5: 67183600-67183600 on Build GRCh38
rs988737596 Chr.5: 67183483-67183483 on Build GRCh38
rs868154912 Chr.5: 67183647-67183647 on Build GRCh38
rs200803485 Chr.5: 67183551-67183551 on Build GRCh38
rs764453962 Chr.5: 67183614-67183614 on Build GRCh38
rs765619451 Chr.5: 67183509-67183509 on Build GRCh38
rs763415618 Chr.5: 67183513-67183513 on Build GRCh38
rs763253387 Chr.5: 67183608-67183608 on Build GRCh38
rs751995367 Chr.5: 67183615-67183615 on Build GRCh38
rs1002487700 Chr.5: 67183527-67183527 on Build GRCh38
rs267600671 Chr.5: 67183505-67183505 on Build GRCh38
rs371897533 Chr.5: 67183552-67183552 on Build GRCh38
rs2230523 Chr.5: 67183555-67183555 on Build GRCh38
rs201238605 Chr.5: 67183454-67183454 on Build GRCh38
rs757302997 Chr.5: 67183533-67183533 on Build GRCh38
rs746951107 Chr.5: 67183561-67183561 on Build GRCh38
rs150317318 Chr.5: 67183622-67183622 on Build GRCh38
rs925777370 Chr.5: 67183449-67183449 on Build GRCh38
rs201686071 Chr.5: 67183646-67183646 on Build GRCh38
rs111374045 Chr.5: 67183605-67183605 on Build GRCh38
rs371692979 Chr.5: 67183487-67183487 on Build GRCh38
rs943218625 Chr.5: 67183576-67183576 on Build GRCh38
rs987708093 Chr.5: 67183559-67183559 on Build GRCh38
rs753105619 Chr.5: 67183511-67183511 on Build GRCh38
rs770467006 Chr.5: 67183570-67183570 on Build GRCh38
rs534148374 Chr.5: 67183617-67183617 on Build GRCh38
rs772176414 Chr.5: 67183447-67183447 on Build GRCh38
rs963686941 Chr.5: 67183548-67183548 on Build GRCh38
rs1025184660 Chr.5: 67183465-67183465 on Build GRCh38
rs750645966 Chr.5: 67183544-67183544 on Build GRCh38
rs772018894 Chr.5: 67183553-67183553 on Build GRCh38
rs750510135 Chr.5: 67183621-67183621 on Build GRCh38
rs568946488 Chr.5: 67183532-67183532 on Build GRCh38
rs776059538 Chr.5: 67183577-67183577 on Build GRCh38
rs200708849 Chr.5: 67183655-67183655 on Build GRCh38
rs137998891 Chr.5: 67183626-67183626 on Build GRCh38
rs891408298 Chr.5: 67183620-67183620 on Build GRCh38
rs756339556 Chr.5: 67183546-67183546 on Build GRCh38
rs762285989 Chr.5: 67183616-67183616 on Build GRCh38
rs776283619 Chr.5: 67183488-67183488 on Build GRCh38
rs1038384169 Chr.5: 67183567-67183567 on Build GRCh38
rs760217507 Chr.5: 67183471-67183471 on Build GRCh38
rs2230522 Chr.5: 67183649-67183649 on Build GRCh38
rs759199601 Chr.5: 67183584-67183584 on Build GRCh38
rs781197790 Chr.5: 67183537-67183537 on Build GRCh38
rs911750251 Chr.5: 67183572-67183572 on Build GRCh38
rs1038904246 Chr.5: 67183601-67183601 on Build GRCh38

View Full Product Details