Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 22347005-22347266 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDR2
Entrez Gene ID
1039
Gene Name
cerebellar degeneration related protein 2
Gene Aliases
CDR62, Yo
UniGene
Hs.513430
Gene Chromosome Location
Chr.16: 22345936-22374617 on Build GRCh38
Transcripts
NM_001802.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5726127 c.1222G>A p.V408I Chr.16: 22347108-22347108 on Build GRCh38
5728353 c.1192C>T p.P398S Chr.16: 22347138-22347138 on Build GRCh38
6486766 c.1126C>T p.H376Y Chr.16: 22347204-22347204 on Build GRCh38
1376723 c.1100G>A p.C367Y Chr.16: 22347230-22347230 on Build GRCh38
7165675 c.1321G>C p.D441H Chr.16: 22347009-22347009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs190419812 Chr.16: 22347172-22347172 on Build GRCh38
rs771570725 Chr.16: 22347203-22347203 on Build GRCh38
rs532368963 Chr.16: 22347127-22347127 on Build GRCh38
rs745338811 Chr.16: 22347196-22347196 on Build GRCh38
rs528530432 Chr.16: 22347186-22347186 on Build GRCh38
rs778041668 Chr.16: 22347183-22347183 on Build GRCh38
rs778387115 Chr.16: 22347081-22347081 on Build GRCh38
rs563641354 Chr.16: 22347126-22347126 on Build GRCh38
rs769832862 Chr.16: 22347233-22347233 on Build GRCh38
rs779592627 Chr.16: 22347206-22347206 on Build GRCh38
rs770429091 Chr.16: 22347062-22347062 on Build GRCh38
rs372824567 Chr.16: 22347058-22347058 on Build GRCh38
rs755150916 Chr.16: 22347040-22347040 on Build GRCh38
rs568142934 Chr.16: 22347228-22347228 on Build GRCh38
rs759852832 Chr.16: 22347135-22347135 on Build GRCh38
rs769961851 Chr.16: 22347157-22347157 on Build GRCh38
rs756495419 Chr.16: 22347178-22347178 on Build GRCh38
rs767606544 Chr.16: 22347147-22347147 on Build GRCh38
rs904981500 Chr.16: 22347075-22347075 on Build GRCh38
rs761900038 Chr.16: 22347232-22347232 on Build GRCh38
rs568513826 Chr.16: 22347238-22347238 on Build GRCh38
rs552181891 Chr.16: 22347129-22347129 on Build GRCh38
rs745588256 Chr.16: 22347093-22347093 on Build GRCh38
rs150205065 Chr.16: 22347185-22347185 on Build GRCh38
rs778440024 Chr.16: 22347190-22347190 on Build GRCh38
rs138712330 Chr.16: 22347210-22347210 on Build GRCh38
rs375939973 Chr.16: 22347236-22347236 on Build GRCh38
rs777255716 Chr.16: 22347059-22347059 on Build GRCh38
rs748238199 Chr.16: 22347104-22347105 on Build GRCh38
rs141072275 Chr.16: 22347094-22347094 on Build GRCh38
rs752986066 Chr.16: 22347051-22347051 on Build GRCh38
rs776564392 Chr.16: 22347222-22347222 on Build GRCh38
rs781278152 Chr.16: 22347099-22347100 on Build GRCh38
rs775394510 Chr.16: 22347097-22347097 on Build GRCh38
rs529904009 Chr.16: 22347046-22347046 on Build GRCh38
rs760418165 Chr.16: 22347121-22347121 on Build GRCh38
rs768520846 Chr.16: 22347125-22347125 on Build GRCh38

View Full Product Details