Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12476905-12477140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP16
Entrez Gene ID
80824
Gene Name
dual specificity phosphatase 16
Gene Aliases
MKP-7, MKP7
UniGene
Hs.536535
Gene Chromosome Location
Chr.12: 12473282-12562839 on Build GRCh38
Transcripts
NM_030640.2 XM_005253488.2 XM_006719155.2 XM_011520856.1 XM_011520857.1 XM_017019988.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
936841 c.1768T>C p.C590R Chr.12: 12477063-12477063 on Build GRCh38
4633553 c.1781T>C p.V594A Chr.12: 12477050-12477050 on Build GRCh38
5728272 c.1715T>C p.I572T Chr.12: 12477116-12477116 on Build GRCh38
5164082 c.1873C>T p.R625C Chr.12: 12476958-12476958 on Build GRCh38
2227576 c.1812T>C p.S604S Chr.12: 12477019-12477019 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750017272 Chr.12: 12477033-12477033 on Build GRCh38
rs866115319 Chr.12: 12476999-12476999 on Build GRCh38
rs763563338 Chr.12: 12477028-12477028 on Build GRCh38
rs746106062 Chr.12: 12477083-12477083 on Build GRCh38
rs759628955 Chr.12: 12477009-12477009 on Build GRCh38
rs529763150 Chr.12: 12477112-12477112 on Build GRCh38
rs778536519 Chr.12: 12476954-12476954 on Build GRCh38
rs145763524 Chr.12: 12477061-12477061 on Build GRCh38
rs941199706 Chr.12: 12477108-12477108 on Build GRCh38
rs748082779 Chr.12: 12476956-12476956 on Build GRCh38
rs778782615 Chr.12: 12477051-12477051 on Build GRCh38
rs147755874 Chr.12: 12477111-12477111 on Build GRCh38
rs758398333 Chr.12: 12477060-12477060 on Build GRCh38
rs562052237 Chr.12: 12477094-12477094 on Build GRCh38
rs771063689 Chr.12: 12477062-12477062 on Build GRCh38
rs775646657 Chr.12: 12477021-12477021 on Build GRCh38
rs754656718 Chr.12: 12477054-12477054 on Build GRCh38
rs753562199 Chr.12: 12477039-12477039 on Build GRCh38
rs770225624 Chr.12: 12477016-12477016 on Build GRCh38
rs144582650 Chr.12: 12476974-12476974 on Build GRCh38
rs532656810 Chr.12: 12476958-12476958 on Build GRCh38
rs202242730 Chr.12: 12477038-12477038 on Build GRCh38
rs201126170 Chr.12: 12477116-12477116 on Build GRCh38
rs867387091 Chr.12: 12477099-12477099 on Build GRCh38
rs771097941 Chr.12: 12477000-12477000 on Build GRCh38
rs781091367 Chr.12: 12477066-12477066 on Build GRCh38
rs150953078 Chr.12: 12477004-12477004 on Build GRCh38
rs771939670 Chr.12: 12476957-12476957 on Build GRCh38
rs754524188 Chr.12: 12477043-12477043 on Build GRCh38
rs764681580 Chr.12: 12477032-12477032 on Build GRCh38
rs1026567580 Chr.12: 12476965-12476965 on Build GRCh38
rs141987644 Chr.12: 12477055-12477055 on Build GRCh38
rs747088797 Chr.12: 12476964-12476964 on Build GRCh38

View Full Product Details