Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 29495857-29496128 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHN2
Entrez Gene ID
1124
Gene Name
chimerin 2
Gene Aliases
ARHGAP3, BCH, CHN2-3, RHOGAP3
UniGene
Hs.654611
Gene Chromosome Location
Chr.7: 29146547-29514335 on Build GRCh38
Transcripts
NM_001039936.2 NM_001293069.1 NM_001293070.1 NM_001293071.1 NM_001293072.1 NM_001293073.1 NM_001293075.1 NM_001293076.1 NM_001293077.1 NM_001293078.1 NM_001293079.1 NM_001293080.1 NM_001293081.1 NM_004067.3 NR_120524.1 NR_120525.1 XM_011515105.2 XM_011515106.2 XM_011515107.2 XM_017011721.1 XM_017011722.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5690871 c.942C>T p.I314I Chr.7: 29496014-29496014 on Build GRCh38
5479477 c.694-8G>T p.? Chr.7: 29495944-29495944 on Build GRCh38
5872564 c.668G>T p.R223L Chr.7: 29495965-29495965 on Build GRCh38
1450276 c.310G>A p.A104T Chr.7: 29496015-29496015 on Build GRCh38
5479475 c.247-8G>T p.? Chr.7: 29495944-29495944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs911434784 Chr.7: 29495913-29495913 on Build GRCh38
rs936809630 Chr.7: 29495879-29495879 on Build GRCh38
rs377591268 Chr.7: 29496028-29496028 on Build GRCh38
rs749358900 Chr.7: 29496067-29496067 on Build GRCh38
rs775497292 Chr.7: 29496064-29496064 on Build GRCh38
rs763140981 Chr.7: 29495981-29495981 on Build GRCh38
rs1001533196 Chr.7: 29496091-29496091 on Build GRCh38
rs777224619 Chr.7: 29495957-29495957 on Build GRCh38
rs780175458 Chr.7: 29495880-29495880 on Build GRCh38
rs751340665 Chr.7: 29495905-29495905 on Build GRCh38
rs751398014 Chr.7: 29495989-29495989 on Build GRCh38
rs746317756 Chr.7: 29496059-29496059 on Build GRCh38
rs760813080 Chr.7: 29495886-29495886 on Build GRCh38
rs748829138 Chr.7: 29495926-29495926 on Build GRCh38
rs765752880 Chr.7: 29496027-29496027 on Build GRCh38
rs750321287 Chr.7: 29496008-29496008 on Build GRCh38
rs781486952 Chr.7: 29495912-29495913 on Build GRCh38
rs761575540 Chr.7: 29496002-29496002 on Build GRCh38
rs779792506 Chr.7: 29495925-29495925 on Build GRCh38
rs200665287 Chr.7: 29496013-29496013 on Build GRCh38
rs146850127 Chr.7: 29496014-29496014 on Build GRCh38
rs764371224 Chr.7: 29495986-29495986 on Build GRCh38
rs3815512 Chr.7: 29496046-29496046 on Build GRCh38
rs755572802 Chr.7: 29495920-29495920 on Build GRCh38
rs771612000 Chr.7: 29495952-29495952 on Build GRCh38
rs761626850 Chr.7: 29496077-29496077 on Build GRCh38
rs767465868 Chr.7: 29496006-29496006 on Build GRCh38
rs772502407 Chr.7: 29495934-29495934 on Build GRCh38
rs372660526 Chr.7: 29496062-29496062 on Build GRCh38
rs913135075 Chr.7: 29496041-29496041 on Build GRCh38
rs545410119 Chr.7: 29496049-29496049 on Build GRCh38
rs775793338 Chr.7: 29495972-29495972 on Build GRCh38
rs745925009 Chr.7: 29496041-29496042 on Build GRCh38
rs757652185 Chr.7: 29496056-29496056 on Build GRCh38
rs749985599 Chr.7: 29495915-29495915 on Build GRCh38
rs756410477 Chr.7: 29496051-29496053 on Build GRCh38
rs555049526 Chr.7: 29495910-29495910 on Build GRCh38
rs184836520 Chr.7: 29495964-29495964 on Build GRCh38
rs778060297 Chr.7: 29495938-29495938 on Build GRCh38
rs146253773 Chr.7: 29496068-29496068 on Build GRCh38
rs757254852 Chr.7: 29495907-29495907 on Build GRCh38
rs759715921 Chr.7: 29495960-29495960 on Build GRCh38
rs767236959 Chr.7: 29496082-29496082 on Build GRCh38
rs774508440 Chr.7: 29496071-29496071 on Build GRCh38
rs781077925 Chr.7: 29495914-29495914 on Build GRCh38
rs149292171 Chr.7: 29495939-29495939 on Build GRCh38

View Full Product Details