Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 25154850-25155103 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf31
Entrez Gene ID
136895
Gene Name
chromosome 7 open reading frame 31
Gene Aliases
-
UniGene
Hs.122055
Gene Chromosome Location
Chr.7: 25134697-25180748 on Build GRCh38
Transcripts
NM_138811.3 XM_005249619.3 XM_005249620.3 XM_011515125.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3950362 c.508T>A p.Y170N Chr.7: 25155098-25155098 on Build GRCh38
6109676 c.513G>C p.T171T Chr.7: 25155093-25155093 on Build GRCh38
3768329 c.585G>T p.L195F Chr.7: 25155021-25155021 on Build GRCh38
5527807 c.544C>T p.P182S Chr.7: 25155062-25155062 on Build GRCh38
5421835 c.512C>T p.T171M Chr.7: 25155094-25155094 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867868119 Chr.7: 25155061-25155062 on Build GRCh38
rs1043731147 Chr.7: 25154875-25154875 on Build GRCh38
rs749786785 Chr.7: 25155040-25155040 on Build GRCh38
rs999649934 Chr.7: 25154909-25154909 on Build GRCh38
rs150213134 Chr.7: 25155079-25155079 on Build GRCh38
rs761960705 Chr.7: 25154969-25154969 on Build GRCh38
rs374602148 Chr.7: 25154972-25154972 on Build GRCh38
rs543319882 Chr.7: 25155018-25155018 on Build GRCh38
rs1024521452 Chr.7: 25155033-25155033 on Build GRCh38
rs373843749 Chr.7: 25155072-25155072 on Build GRCh38
rs145737599 Chr.7: 25155011-25155011 on Build GRCh38
rs534391492 Chr.7: 25155008-25155008 on Build GRCh38
rs753260167 Chr.7: 25154994-25154994 on Build GRCh38
rs386711396 Chr.7: 25155017-25155018 on Build GRCh38
rs756665589 Chr.7: 25155029-25155029 on Build GRCh38
rs762608903 Chr.7: 25155062-25155063 on Build GRCh38
rs759406357 Chr.7: 25155076-25155076 on Build GRCh38
rs368042746 Chr.7: 25155080-25155080 on Build GRCh38
rs528491270 Chr.7: 25154945-25154945 on Build GRCh38
rs201143588 Chr.7: 25154968-25154968 on Build GRCh38
rs772340513 Chr.7: 25155049-25155049 on Build GRCh38
rs766131857 Chr.7: 25154995-25154995 on Build GRCh38
rs370479494 Chr.7: 25155055-25155055 on Build GRCh38
rs896322161 Chr.7: 25154952-25154952 on Build GRCh38
rs149091804 Chr.7: 25155017-25155017 on Build GRCh38
rs760596307 Chr.7: 25154984-25154984 on Build GRCh38
rs766355678 Chr.7: 25154989-25154989 on Build GRCh38
rs539268352 Chr.7: 25154899-25154899 on Build GRCh38
rs988923472 Chr.7: 25155067-25155067 on Build GRCh38
rs984435272 Chr.7: 25154997-25154997 on Build GRCh38
rs775563238 Chr.7: 25155014-25155015 on Build GRCh38
rs12538022 Chr.7: 25154887-25154887 on Build GRCh38
rs371193551 Chr.7: 25154970-25154970 on Build GRCh38
rs752338617 Chr.7: 25155010-25155010 on Build GRCh38
rs1013473857 Chr.7: 25154953-25154953 on Build GRCh38
rs2523073 Chr.7: 25154999-25154999 on Build GRCh38
rs200527864 Chr.7: 25155068-25155068 on Build GRCh38
rs557172191 Chr.7: 25155037-25155037 on Build GRCh38
rs768728755 Chr.7: 25155043-25155043 on Build GRCh38
rs117283904 Chr.7: 25155066-25155066 on Build GRCh38
rs906035305 Chr.7: 25155039-25155039 on Build GRCh38
rs141468448 Chr.7: 25155062-25155062 on Build GRCh38
rs2523072 Chr.7: 25155046-25155046 on Build GRCh38

View Full Product Details