Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45616338-45616597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML2
Entrez Gene ID
24139
Gene Name
echinoderm microtubule associated protein like 2
Gene Aliases
ELP70, EMAP-2, EMAP2
UniGene
Hs.24178
Gene Chromosome Location
Chr.19: 45609396-45645601 on Build GRCh38
Transcripts
NM_001193268.1 NM_001193269.1 NM_012155.2 NR_034098.1 XM_006723125.2 XM_006723126.2 XM_011526694.2 XM_011526695.2 XM_011526698.2 XM_011526700.2 XM_011526701.2 XM_017026550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6653760 c.1853-3delC p.? Chr.19: 45616561-45616561 on Build GRCh38
6653750 c.2044G>A p.D682N Chr.19: 45616529-45616529 on Build GRCh38
4891804 c.2039C>T p.S680F Chr.19: 45616534-45616534 on Build GRCh38
5474183 c.1482C>G p.R494R Chr.19: 45616488-45616488 on Build GRCh38
6653749 c.1882G>A p.D628N Chr.19: 45616529-45616529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764647472 Chr.19: 45616424-45616424 on Build GRCh38
rs199584109 Chr.19: 45616514-45616514 on Build GRCh38
rs1050655021 Chr.19: 45616388-45616388 on Build GRCh38
rs771030163 Chr.19: 45616465-45616465 on Build GRCh38
rs562531317 Chr.19: 45616379-45616379 on Build GRCh38
rs752102640 Chr.19: 45616543-45616543 on Build GRCh38
rs748316353 Chr.19: 45616438-45616438 on Build GRCh38
rs766939226 Chr.19: 45616430-45616430 on Build GRCh38
rs777873246 Chr.19: 45616461-45616461 on Build GRCh38
rs749341374 Chr.19: 45616463-45616463 on Build GRCh38
rs753817789 Chr.19: 45616498-45616498 on Build GRCh38
rs902110528 Chr.19: 45616506-45616506 on Build GRCh38
rs1052041810 Chr.19: 45616374-45616374 on Build GRCh38
rs938989949 Chr.19: 45616392-45616392 on Build GRCh38
rs757857153 Chr.19: 45616429-45616429 on Build GRCh38
rs778018008 Chr.19: 45616578-45616578 on Build GRCh38
rs1008165335 Chr.19: 45616477-45616477 on Build GRCh38
rs765256711 Chr.19: 45616572-45616573 on Build GRCh38
rs761517168 Chr.19: 45616495-45616495 on Build GRCh38
rs866166593 Chr.19: 45616489-45616489 on Build GRCh38
rs768145377 Chr.19: 45616564-45616564 on Build GRCh38
rs755612839 Chr.19: 45616554-45616554 on Build GRCh38
rs111418701 Chr.19: 45616389-45616389 on Build GRCh38
rs373983729 Chr.19: 45616555-45616555 on Build GRCh38
rs765878588 Chr.19: 45616431-45616431 on Build GRCh38
rs759767127 Chr.19: 45616484-45616495 on Build GRCh38
rs376382973 Chr.19: 45616378-45616378 on Build GRCh38
rs761584217 Chr.19: 45616422-45616422 on Build GRCh38
rs756458089 Chr.19: 45616568-45616568 on Build GRCh38
rs533694573 Chr.19: 45616524-45616524 on Build GRCh38
rs551493246 Chr.19: 45616478-45616478 on Build GRCh38
rs956286309 Chr.19: 45616573-45616573 on Build GRCh38
rs376045352 Chr.19: 45616470-45616470 on Build GRCh38
rs918967228 Chr.19: 45616423-45616423 on Build GRCh38
rs753465919 Chr.19: 45616412-45616412 on Build GRCh38
rs749878867 Chr.19: 45616428-45616428 on Build GRCh38
rs370545238 Chr.19: 45616507-45616507 on Build GRCh38
rs781730329 Chr.19: 45616436-45616436 on Build GRCh38
rs756296035 Chr.19: 45616459-45616459 on Build GRCh38
rs866961255 Chr.19: 45616462-45616462 on Build GRCh38
rs1056074114 Chr.19: 45616394-45616394 on Build GRCh38
rs766022510 Chr.19: 45616542-45616542 on Build GRCh38
rs762480985 Chr.19: 45616515-45616515 on Build GRCh38
rs547198368 Chr.19: 45616532-45616532 on Build GRCh38
rs753050205 Chr.19: 45616565-45616565 on Build GRCh38
rs993631724 Chr.19: 45616525-45616525 on Build GRCh38

View Full Product Details