Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 70816504-70816761 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4F
Entrez Gene ID
165530
Gene Name
C-type lectin domain family 4 member F
Gene Aliases
CLECSF13, KCLR
UniGene
Hs.681239
Gene Chromosome Location
Chr.2: 70808643-70825235 on Build GRCh38
Transcripts
NM_001258027.1 NM_001321308.1 NM_173535.2 XM_011532635.2 XM_011532637.2 XM_011532638.2 XM_011532639.2 XM_011532640.2 XM_011532641.2 XM_011532642.2 XM_011532643.1 XM_017003519.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3695578 c.692C>T p.A231V Chr.2: 70816689-70816689 on Build GRCh38
3702222 c.694A>T p.S232C Chr.2: 70816687-70816687 on Build GRCh38
1742504 c.704C>T p.T235M Chr.2: 70816677-70816677 on Build GRCh38
1022542 c.712A>G p.T238A Chr.2: 70816669-70816669 on Build GRCh38
1022543 c.640G>A p.V214M Chr.2: 70816741-70816741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781804335 Chr.2: 70816622-70816622 on Build GRCh38
rs781822853 Chr.2: 70816633-70816633 on Build GRCh38
rs781927860 Chr.2: 70816568-70816568 on Build GRCh38
rs375785739 Chr.2: 70816635-70816635 on Build GRCh38
rs782190255 Chr.2: 70816687-70816687 on Build GRCh38
rs139690383 Chr.2: 70816639-70816639 on Build GRCh38
rs370818847 Chr.2: 70816676-70816676 on Build GRCh38
rs146602856 Chr.2: 70816698-70816698 on Build GRCh38
rs150218766 Chr.2: 70816741-70816741 on Build GRCh38
rs1033411670 Chr.2: 70816590-70816590 on Build GRCh38
rs782010298 Chr.2: 70816681-70816681 on Build GRCh38
rs782047460 Chr.2: 70816623-70816623 on Build GRCh38
rs1022093869 Chr.2: 70816540-70816540 on Build GRCh38
rs781810783 Chr.2: 70816739-70816739 on Build GRCh38
rs782778887 Chr.2: 70816717-70816717 on Build GRCh38
rs782154444 Chr.2: 70816609-70816609 on Build GRCh38
rs782304657 Chr.2: 70816689-70816689 on Build GRCh38
rs62622841 Chr.2: 70816630-70816630 on Build GRCh38
rs781969025 Chr.2: 70816697-70816697 on Build GRCh38
rs78458500 Chr.2: 70816677-70816677 on Build GRCh38
rs782170151 Chr.2: 70816731-70816731 on Build GRCh38
rs373834534 Chr.2: 70816536-70816536 on Build GRCh38
rs782278242 Chr.2: 70816537-70816537 on Build GRCh38
rs782331467 Chr.2: 70816549-70816549 on Build GRCh38
rs782804774 Chr.2: 70816621-70816621 on Build GRCh38
rs142908853 Chr.2: 70816634-70816634 on Build GRCh38
rs781978886 Chr.2: 70816601-70816601 on Build GRCh38
rs782610041 Chr.2: 70816673-70816673 on Build GRCh38
rs376668351 Chr.2: 70816672-70816672 on Build GRCh38
rs113852847 Chr.2: 70816649-70816649 on Build GRCh38
rs371728170 Chr.2: 70816742-70816742 on Build GRCh38
rs367726937 Chr.2: 70816686-70816686 on Build GRCh38
rs78241995 Chr.2: 70816596-70816596 on Build GRCh38
rs142040724 Chr.2: 70816652-70816652 on Build GRCh38
rs782531692 Chr.2: 70816645-70816645 on Build GRCh38
rs1001931433 Chr.2: 70816569-70816569 on Build GRCh38
rs782810666 Chr.2: 70816738-70816738 on Build GRCh38
rs782108527 Chr.2: 70816577-70816577 on Build GRCh38
rs200019956 Chr.2: 70816719-70816719 on Build GRCh38

View Full Product Details