Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 92776791-92777020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EEA1
Entrez Gene ID
8411
Gene Name
early endosome antigen 1
Gene Aliases
MST105, MSTP105, ZFYVE2
UniGene
Hs.567367
Gene Chromosome Location
Chr.12: 92772509-92975228 on Build GRCh38
Transcripts
NM_003566.3 XM_011538814.2 XM_011538815.2 XM_017020018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6323541 c.4040G>A p.R1347K Chr.12: 92776917-92776917 on Build GRCh38
4045407 c.4111C>T p.R1371W Chr.12: 92776846-92776846 on Build GRCh38
4935869 c.4100T>C p.V1367A Chr.12: 92776857-92776857 on Build GRCh38
4045409 c.4050C>T p.A1350A Chr.12: 92776907-92776907 on Build GRCh38
6239356 c.4015-1G>T p.? Chr.12: 92776943-92776943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775693118 Chr.12: 92776987-92776987 on Build GRCh38
rs35262051 Chr.12: 92776861-92776861 on Build GRCh38
rs750282542 Chr.12: 92776874-92776874 on Build GRCh38
rs767471657 Chr.12: 92776976-92776976 on Build GRCh38
rs759709731 Chr.12: 92776845-92776845 on Build GRCh38
rs766714962 Chr.12: 92776839-92776839 on Build GRCh38
rs774743190 Chr.12: 92776943-92776943 on Build GRCh38
rs773647272 Chr.12: 92776932-92776932 on Build GRCh38
rs757264285 Chr.12: 92776857-92776857 on Build GRCh38
rs560597630 Chr.12: 92776846-92776846 on Build GRCh38
rs1029930428 Chr.12: 92776963-92776963 on Build GRCh38
rs772933667 Chr.12: 92776853-92776853 on Build GRCh38
rs759842673 Chr.12: 92776962-92776962 on Build GRCh38
rs780807493 Chr.12: 92776923-92776923 on Build GRCh38
rs778555063 Chr.12: 92776867-92776867 on Build GRCh38
rs142495165 Chr.12: 92776907-92776907 on Build GRCh38
rs763265815 Chr.12: 92776826-92776826 on Build GRCh38
rs757976471 Chr.12: 92776882-92776882 on Build GRCh38
rs770399482 Chr.12: 92776931-92776931 on Build GRCh38
rs753686694 Chr.12: 92776856-92776856 on Build GRCh38
rs1038290258 Chr.12: 92776933-92776933 on Build GRCh38
rs748590154 Chr.12: 92776927-92776927 on Build GRCh38
rs771227792 Chr.12: 92776934-92776934 on Build GRCh38
rs779706031 Chr.12: 92776889-92776889 on Build GRCh38
rs751630827 Chr.12: 92776840-92776840 on Build GRCh38
rs754419885 Chr.12: 92776921-92776921 on Build GRCh38

View Full Product Details