Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 95535077-95535320 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRX5
Entrez Gene ID
51218
Gene Name
glutaredoxin 5
Gene Aliases
C14orf87, FLB4739, GRX5, PR01238, PRO1238, PRSA, SIDBA3, SPAHGC
UniGene
Hs.744943
Gene Chromosome Location
Chr.14: 95534986-95544718 on Build GRCh38
Transcripts
NM_016417.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7328657 c.49C>A p.R17S Chr.14: 95535138-95535138 on Build GRCh38
4649533 c.212A>G p.N71S Chr.14: 95535301-95535301 on Build GRCh38
7328493 c.37C>A p.L13I Chr.14: 95535126-95535126 on Build GRCh38
5688444 c.106G>T p.A36S Chr.14: 95535195-95535195 on Build GRCh38
5678429 c.19C>G p.R7G Chr.14: 95535108-95535108 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs972849547 Chr.14: 95535179-95535200 on Build GRCh38
rs998957812 Chr.14: 95535250-95535250 on Build GRCh38
rs747458426 Chr.14: 95535240-95535240 on Build GRCh38
rs587776500 Chr.14: 95535218-95535218 on Build GRCh38
rs1027227294 Chr.14: 95535296-95535296 on Build GRCh38
rs558960545 Chr.14: 95535269-95535269 on Build GRCh38
rs573438674 Chr.14: 95535103-95535103 on Build GRCh38
rs990377177 Chr.14: 95535222-95535222 on Build GRCh38
rs758899397 Chr.14: 95535223-95535223 on Build GRCh38
rs753486590 Chr.14: 95535190-95535190 on Build GRCh38
rs760361132 Chr.14: 95535112-95535112 on Build GRCh38
rs894818908 Chr.14: 95535098-95535098 on Build GRCh38
rs534195488 Chr.14: 95535204-95535204 on Build GRCh38
rs577438813 Chr.14: 95535274-95535274 on Build GRCh38
rs765749786 Chr.14: 95535176-95535194 on Build GRCh38
rs869320757 Chr.14: 95535240-95535242 on Build GRCh38
rs888641515 Chr.14: 95535297-95535297 on Build GRCh38
rs964398307 Chr.14: 95535221-95535221 on Build GRCh38
rs750771696 Chr.14: 95535197-95535198 on Build GRCh38
rs746028473 Chr.14: 95535260-95535260 on Build GRCh38
rs7158346 Chr.14: 95535272-95535272 on Build GRCh38
rs869320758 Chr.14: 95535182-95535183 on Build GRCh38
rs1008334686 Chr.14: 95535197-95535197 on Build GRCh38
rs781621057 Chr.14: 95535257-95535257 on Build GRCh38
rs778023417 Chr.14: 95535227-95535227 on Build GRCh38
rs898452545 Chr.14: 95535152-95535152 on Build GRCh38
rs993992614 Chr.14: 95535168-95535168 on Build GRCh38
rs757690699 Chr.14: 95535243-95535243 on Build GRCh38
rs955421878 Chr.14: 95535195-95535195 on Build GRCh38
rs375714975 Chr.14: 95535130-95535130 on Build GRCh38
rs767242677 Chr.14: 95535096-95535096 on Build GRCh38
rs71431696 Chr.14: 95535293-95535293 on Build GRCh38
rs895229563 Chr.14: 95535256-95535256 on Build GRCh38
rs766056735 Chr.14: 95535185-95535185 on Build GRCh38
rs754544048 Chr.14: 95535201-95535201 on Build GRCh38
rs760533176 Chr.14: 95535302-95535302 on Build GRCh38
rs753153633 Chr.14: 95535212-95535212 on Build GRCh38
rs748054600 Chr.14: 95535282-95535282 on Build GRCh38
rs28680410 Chr.14: 95535287-95535287 on Build GRCh38

View Full Product Details