Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 58492138-58492399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP7A1
Entrez Gene ID
1581
Gene Name
cytochrome P450 family 7 subfamily A member 1
Gene Aliases
CP7A, CYP7, CYPVII
UniGene
Hs.1644
Gene Chromosome Location
Chr.8: 58490178-58500161 on Build GRCh38
Transcripts
NM_000780.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1100715 c.1203C>T p.Y401Y Chr.8: 58492365-58492365 on Build GRCh38
7236117 c.1213T>C p.L405L Chr.8: 58492355-58492355 on Build GRCh38
6181229 c.1183C>A p.H395N Chr.8: 58492385-58492385 on Build GRCh38
7173511 c.1210C>T p.P404S Chr.8: 58492358-58492358 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1051568434 Chr.8: 58492278-58492278 on Build GRCh38
rs867871046 Chr.8: 58492329-58492329 on Build GRCh38
rs908617307 Chr.8: 58492285-58492285 on Build GRCh38
rs900107676 Chr.8: 58492258-58492258 on Build GRCh38
rs746854399 Chr.8: 58492312-58492312 on Build GRCh38
rs375383265 Chr.8: 58492337-58492337 on Build GRCh38
rs368417648 Chr.8: 58492345-58492345 on Build GRCh38
rs931133038 Chr.8: 58492249-58492249 on Build GRCh38
rs140910170 Chr.8: 58492189-58492189 on Build GRCh38
rs910018528 Chr.8: 58492232-58492232 on Build GRCh38
rs1012727318 Chr.8: 58492372-58492372 on Build GRCh38
rs199664989 Chr.8: 58492319-58492319 on Build GRCh38
rs568264638 Chr.8: 58492281-58492281 on Build GRCh38
rs552145192 Chr.8: 58492197-58492197 on Build GRCh38
rs965014864 Chr.8: 58492202-58492202 on Build GRCh38
rs912252931 Chr.8: 58492192-58492192 on Build GRCh38
rs780054084 Chr.8: 58492310-58492310 on Build GRCh38
rs753377633 Chr.8: 58492355-58492355 on Build GRCh38
rs77341564 Chr.8: 58492268-58492268 on Build GRCh38
rs761084132 Chr.8: 58492353-58492353 on Build GRCh38
rs372356911 Chr.8: 58492321-58492321 on Build GRCh38
rs372341079 Chr.8: 58492354-58492354 on Build GRCh38
rs903265207 Chr.8: 58492279-58492279 on Build GRCh38
rs776072887 Chr.8: 58492350-58492350 on Build GRCh38
rs759920039 Chr.8: 58492341-58492341 on Build GRCh38
rs771107553 Chr.8: 58492324-58492324 on Build GRCh38
rs569057537 Chr.8: 58492221-58492221 on Build GRCh38
rs941525413 Chr.8: 58492240-58492240 on Build GRCh38
rs769980711 Chr.8: 58492318-58492318 on Build GRCh38
rs779251933 Chr.8: 58492364-58492364 on Build GRCh38
rs531837289 Chr.8: 58492245-58492245 on Build GRCh38

View Full Product Details