Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68230850-68231101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRP2
Entrez Gene ID
80004
Gene Name
epithelial splicing regulatory protein 2
Gene Aliases
RBM35B
UniGene
Hs.436585
Gene Chromosome Location
Chr.16: 68228547-68236233 on Build GRCh38
Transcripts
NM_024939.2 XM_005256153.4 XM_006721273.3 XM_011523338.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2996300 c.1754C>T p.T585M Chr.16: 68230985-68230985 on Build GRCh38
4883651 c.1736C>A p.T579N Chr.16: 68231003-68231003 on Build GRCh38
5485815 c.1712-1G>A p.? Chr.16: 68231028-68231028 on Build GRCh38
7366653 c.1784C>T p.T595M Chr.16: 68230985-68230985 on Build GRCh38
7101048 c.1788delC p.S597fs*29 Chr.16: 68230951-68230951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748388857 Chr.16: 68230888-68230888 on Build GRCh38
rs754700540 Chr.16: 68230903-68230904 on Build GRCh38
rs556537582 Chr.16: 68230967-68230967 on Build GRCh38
rs756906275 Chr.16: 68230948-68230948 on Build GRCh38
rs763972371 Chr.16: 68230938-68230938 on Build GRCh38
rs762966945 Chr.16: 68231020-68231020 on Build GRCh38
rs1046606817 Chr.16: 68230989-68230989 on Build GRCh38
rs117260313 Chr.16: 68230951-68230951 on Build GRCh38
rs749792127 Chr.16: 68230972-68230972 on Build GRCh38
rs768871334 Chr.16: 68230885-68230885 on Build GRCh38
rs759422467 Chr.16: 68230909-68230909 on Build GRCh38
rs755446587 Chr.16: 68231058-68231058 on Build GRCh38
rs762766253 Chr.16: 68230928-68230928 on Build GRCh38
rs756594868 Chr.16: 68230966-68230966 on Build GRCh38
rs200279617 Chr.16: 68230912-68230912 on Build GRCh38
rs779280493 Chr.16: 68231059-68231059 on Build GRCh38
rs772296415 Chr.16: 68230988-68230988 on Build GRCh38
rs537836223 Chr.16: 68230954-68230954 on Build GRCh38
rs755718291 Chr.16: 68231041-68231041 on Build GRCh38
rs748358961 Chr.16: 68230985-68230985 on Build GRCh38
rs111882305 Chr.16: 68231081-68231081 on Build GRCh38
rs756606781 Chr.16: 68230876-68230876 on Build GRCh38
rs377474438 Chr.16: 68230979-68230979 on Build GRCh38
rs775436384 Chr.16: 68230920-68230920 on Build GRCh38
rs201684281 Chr.16: 68231035-68231035 on Build GRCh38
rs755393677 Chr.16: 68230973-68230973 on Build GRCh38
rs888716792 Chr.16: 68231043-68231043 on Build GRCh38
rs951860481 Chr.16: 68230891-68230891 on Build GRCh38
rs375171498 Chr.16: 68231036-68231036 on Build GRCh38
rs199808836 Chr.16: 68231030-68231030 on Build GRCh38
rs773185656 Chr.16: 68230995-68230995 on Build GRCh38
rs552251668 Chr.16: 68230941-68230941 on Build GRCh38
rs753055981 Chr.16: 68231063-68231063 on Build GRCh38
rs749599581 Chr.16: 68230877-68230877 on Build GRCh38
rs761396911 Chr.16: 68231034-68231034 on Build GRCh38
rs192768082 Chr.16: 68231044-68231044 on Build GRCh38
rs145134851 Chr.16: 68231003-68231003 on Build GRCh38
rs775632478 Chr.16: 68231010-68231010 on Build GRCh38
rs773310955 Chr.16: 68230905-68230905 on Build GRCh38
rs754392299 Chr.16: 68231049-68231049 on Build GRCh38
rs36054935 Chr.16: 68230890-68230890 on Build GRCh38
rs774252203 Chr.16: 68231033-68231033 on Build GRCh38
rs200397589 Chr.16: 68231008-68231008 on Build GRCh38
rs752592064 Chr.16: 68231032-68231032 on Build GRCh38

View Full Product Details