Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 48953695-48953944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARFGEF2
Entrez Gene ID
10564
Gene Name
ADP ribosylation factor guanine nucleotide exchange factor 2
Gene Aliases
BIG2, PVNH2, dJ1164I10.1
UniGene
Hs.62578
Gene Chromosome Location
Chr.20: 48921721-49036693 on Build GRCh38
Transcripts
NM_006420.2 XM_005260252.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1027663 c.770G>T p.R257M Chr.20: 48953722-48953722 on Build GRCh38
4850997 c.822A>T p.R274S Chr.20: 48953774-48953774 on Build GRCh38
1027665 c.808G>A p.A270T Chr.20: 48953760-48953760 on Build GRCh38
5896422 c.779C>T p.S260F Chr.20: 48953731-48953731 on Build GRCh38
3547450 c.813C>T p.P271P Chr.20: 48953765-48953765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778834589 Chr.20: 48953760-48953760 on Build GRCh38
rs751655373 Chr.20: 48953741-48953741 on Build GRCh38
rs143570842 Chr.20: 48953763-48953763 on Build GRCh38
rs752907103 Chr.20: 48953839-48953839 on Build GRCh38
rs373985686 Chr.20: 48953804-48953804 on Build GRCh38
rs766629171 Chr.20: 48953729-48953729 on Build GRCh38
rs938736539 Chr.20: 48953762-48953762 on Build GRCh38
rs781424305 Chr.20: 48953746-48953746 on Build GRCh38
rs776283531 Chr.20: 48953777-48953777 on Build GRCh38
rs767636886 Chr.20: 48953821-48953821 on Build GRCh38
rs370588776 Chr.20: 48953786-48953786 on Build GRCh38
rs145245951 Chr.20: 48953758-48953758 on Build GRCh38
rs748477785 Chr.20: 48953784-48953784 on Build GRCh38
rs533047547 Chr.20: 48953776-48953776 on Build GRCh38
rs746481367 Chr.20: 48953769-48953769 on Build GRCh38
rs530687189 Chr.20: 48953844-48953844 on Build GRCh38
rs1017215376 Chr.20: 48953871-48953871 on Build GRCh38
rs560939541 Chr.20: 48953889-48953889 on Build GRCh38
rs749160098 Chr.20: 48953862-48953862 on Build GRCh38
rs375582617 Chr.20: 48953767-48953767 on Build GRCh38
rs961436174 Chr.20: 48953883-48953883 on Build GRCh38
rs149172723 Chr.20: 48953759-48953759 on Build GRCh38
rs755201684 Chr.20: 48953743-48953743 on Build GRCh38
rs773712651 Chr.20: 48953794-48953794 on Build GRCh38
rs150046458 Chr.20: 48953722-48953722 on Build GRCh38
rs1043533503 Chr.20: 48953766-48953766 on Build GRCh38
rs763591309 Chr.20: 48953800-48953800 on Build GRCh38
rs767149566 Chr.20: 48953725-48953725 on Build GRCh38
rs368437314 Chr.20: 48953805-48953805 on Build GRCh38
rs974126758 Chr.20: 48953898-48953898 on Build GRCh38
rs569846581 Chr.20: 48953842-48953842 on Build GRCh38
rs1027052120 Chr.20: 48953914-48953914 on Build GRCh38
rs997198981 Chr.20: 48953920-48953920 on Build GRCh38

View Full Product Details