Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13654351-13654594 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD4A
Entrez Gene ID
55691
Gene Name
FERM domain containing 4A
Gene Aliases
CCAFCA, FRMD4, bA295P9.4
UniGene
Hs.330463
Gene Chromosome Location
Chr.10: 13643706-14330952 on Build GRCh38
Transcripts
NM_001318336.1 NM_001318337.1 NM_001318338.1 NM_018027.4 XM_005252489.2 XM_005252490.2 XM_005252495.4 XM_006717457.2 XM_006717458.3 XM_006717459.2 XM_006717460.2 XM_006717464.1 XM_011519539.1 XM_011519540.1 XM_011519541.1 XM_011519543.2 XM_017016393.1 XM_017016394.1 XM_017016395.1 XM_017016396.1

Gene Symbol
LOC101928524
Entrez Gene ID
101928524
Gene Name
uncharacterized LOC101928524
Gene Aliases
-
UniGene
Hs.161181
Gene Chromosome Location
Chr.10: 13650927-13655929 on Build GRCh38
Transcripts
NM_001318336.1 NM_001318337.1 NM_001318338.1 NM_018027.4 XM_005252489.2 XM_005252490.2 XM_005252495.4 XM_006717457.2 XM_006717458.3 XM_006717459.2 XM_006717460.2 XM_006717464.1 XM_011519539.1 XM_011519540.1 XM_011519541.1 XM_011519543.2 XM_017016393.1 XM_017016394.1 XM_017016395.1 XM_017016396.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1346988 c.3014delC p.P1005fs*8 Chr.10: 13654452-13654452 on Build GRCh38
4804815 c.2954C>A p.A985D Chr.10: 13654512-13654512 on Build GRCh38
1685657 c.3014_3015insC p.S1006fs*25 Chr.10: 13654451-13654452 on Build GRCh38
4765630 c.3015_3016insC p.S1006fs*25 Chr.10: 13654450-13654451 on Build GRCh38
4486111 c.3012C>T p.P1004P Chr.10: 13654454-13654454 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756413355 Chr.10: 13654529-13654529 on Build GRCh38
rs760585112 Chr.10: 13654434-13654434 on Build GRCh38
rs10906442 Chr.10: 13654570-13654570 on Build GRCh38
rs764804844 Chr.10: 13654448-13654448 on Build GRCh38
rs752115977 Chr.10: 13654386-13654386 on Build GRCh38
rs199934183 Chr.10: 13654479-13654479 on Build GRCh38
rs1048169983 Chr.10: 13654423-13654423 on Build GRCh38
rs764859930 Chr.10: 13654510-13654510 on Build GRCh38
rs760389029 Chr.10: 13654377-13654377 on Build GRCh38
rs754948936 Chr.10: 13654404-13654406 on Build GRCh38
rs755204978 Chr.10: 13654551-13654551 on Build GRCh38
rs149301184 Chr.10: 13654504-13654504 on Build GRCh38
rs781510353 Chr.10: 13654539-13654540 on Build GRCh38
rs931037024 Chr.10: 13654402-13654402 on Build GRCh38
rs373403491 Chr.10: 13654399-13654399 on Build GRCh38
rs114708314 Chr.10: 13654378-13654378 on Build GRCh38
rs535596804 Chr.10: 13654450-13654450 on Build GRCh38
rs780203300 Chr.10: 13654460-13654460 on Build GRCh38
rs778915941 Chr.10: 13654414-13654415 on Build GRCh38
rs377693430 Chr.10: 13654478-13654478 on Build GRCh38
rs570069245 Chr.10: 13654571-13654571 on Build GRCh38
rs776441796 Chr.10: 13654441-13654441 on Build GRCh38
rs748112574 Chr.10: 13654429-13654429 on Build GRCh38
rs769984841 Chr.10: 13654416-13654416 on Build GRCh38
rs918800644 Chr.10: 13654394-13654394 on Build GRCh38
rs768746476 Chr.10: 13654426-13654426 on Build GRCh38
rs1040733760 Chr.10: 13654469-13654469 on Build GRCh38
rs768029606 Chr.10: 13654522-13654522 on Build GRCh38
rs752538869 Chr.10: 13654451-13654452 on Build GRCh38
rs754984701 Chr.10: 13654465-13654465 on Build GRCh38
rs771960572 Chr.10: 13654430-13654430 on Build GRCh38
rs1035686769 Chr.10: 13654573-13654573 on Build GRCh38
rs1011170629 Chr.10: 13654486-13654486 on Build GRCh38
rs748217128 Chr.10: 13654556-13654556 on Build GRCh38
rs756503357 Chr.10: 13654454-13654454 on Build GRCh38
rs537063210 Chr.10: 13654542-13654542 on Build GRCh38
rs779037775 Chr.10: 13654467-13654467 on Build GRCh38
rs138561619 Chr.10: 13654425-13654425 on Build GRCh38
rs374192557 Chr.10: 13654484-13654484 on Build GRCh38
rs568514807 Chr.10: 13654485-13654485 on Build GRCh38
rs377377541 Chr.10: 13654400-13654400 on Build GRCh38
rs778089059 Chr.10: 13654385-13654385 on Build GRCh38
rs368934904 Chr.10: 13654427-13654427 on Build GRCh38
rs374464932 Chr.10: 13654453-13654453 on Build GRCh38
rs758083736 Chr.10: 13654478-13654479 on Build GRCh38
rs780296999 Chr.10: 13654422-13654422 on Build GRCh38
rs377357288 Chr.10: 13654517-13654517 on Build GRCh38
rs369209502 Chr.10: 13654392-13654392 on Build GRCh38
rs1017080215 Chr.10: 13654499-13654499 on Build GRCh38
rs990652950 Chr.10: 13654452-13654452 on Build GRCh38
rs750952078 Chr.10: 13654528-13654528 on Build GRCh38
rs762412813 Chr.10: 13654519-13654519 on Build GRCh38
rs1003935115 Chr.10: 13654498-13654498 on Build GRCh38
rs779041591 Chr.10: 13654554-13654554 on Build GRCh38
rs754257759 Chr.10: 13654546-13654546 on Build GRCh38
rs753871837 Chr.10: 13654401-13654403 on Build GRCh38
rs759054988 Chr.10: 13654446-13654446 on Build GRCh38
rs749526008 Chr.10: 13654462-13654462 on Build GRCh38
rs754563506 Chr.10: 13654379-13654379 on Build GRCh38
rs773036672 Chr.10: 13654432-13654432 on Build GRCh38

View Full Product Details