Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207724605-207724876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNYL1
Entrez Gene ID
151195
Gene Name
cyclin Y like 1
Gene Aliases
-
UniGene
Hs.471234
Gene Chromosome Location
Chr.2: 207711540-207756174 on Build GRCh38
Transcripts
NM_001142300.1 NM_152523.2 XM_005246343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2714473 c.60T>G p.I20M Chr.2: 207724849-207724849 on Build GRCh38
6862044 c.284C>G p.S95C Chr.2: 207724863-207724863 on Build GRCh38
2714472 c.270T>G p.I90M Chr.2: 207724849-207724849 on Build GRCh38
3838428 c.253C>T p.P85S Chr.2: 207724832-207724832 on Build GRCh38
3838431 c.76C>G p.Q26E Chr.2: 207724865-207724865 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771127011 Chr.2: 207724757-207724757 on Build GRCh38
rs888997931 Chr.2: 207724628-207724628 on Build GRCh38
rs748491839 Chr.2: 207724838-207724838 on Build GRCh38
rs753471957 Chr.2: 207724810-207724810 on Build GRCh38
rs936604672 Chr.2: 207724647-207724647 on Build GRCh38
rs74570049 Chr.2: 207724626-207724626 on Build GRCh38
rs572361324 Chr.2: 207724659-207724659 on Build GRCh38
rs1034355559 Chr.2: 207724786-207724786 on Build GRCh38
rs772492989 Chr.2: 207724752-207724752 on Build GRCh38
rs751450607 Chr.2: 207724767-207724767 on Build GRCh38
rs779602983 Chr.2: 207724789-207724789 on Build GRCh38
rs760690641 Chr.2: 207724754-207724754 on Build GRCh38
rs1052497937 Chr.2: 207724835-207724835 on Build GRCh38
rs772627737 Chr.2: 207724651-207724651 on Build GRCh38
rs763968055 Chr.2: 207724760-207724760 on Build GRCh38
rs182255224 Chr.2: 207724679-207724679 on Build GRCh38
rs780799169 Chr.2: 207724789-207724793 on Build GRCh38
rs372038496 Chr.2: 207724769-207724769 on Build GRCh38
rs959187149 Chr.2: 207724803-207724803 on Build GRCh38
rs925245416 Chr.2: 207724796-207724796 on Build GRCh38
rs962449933 Chr.2: 207724648-207724648 on Build GRCh38
rs892290598 Chr.2: 207724689-207724689 on Build GRCh38
rs951547346 Chr.2: 207724730-207724730 on Build GRCh38
rs759904707 Chr.2: 207724759-207724759 on Build GRCh38
rs767389524 Chr.2: 207724776-207724776 on Build GRCh38
rs754672700 Chr.2: 207724813-207724813 on Build GRCh38
rs760180232 Chr.2: 207724738-207724738 on Build GRCh38
rs60537915 Chr.2: 207724778-207724778 on Build GRCh38
rs1009425964 Chr.2: 207724697-207724697 on Build GRCh38
rs779215198 Chr.2: 207724814-207724814 on Build GRCh38
rs866480151 Chr.2: 207724847-207724847 on Build GRCh38
rs201730087 Chr.2: 207724753-207724753 on Build GRCh38
rs185205884 Chr.2: 207724788-207724788 on Build GRCh38
rs749903477 Chr.2: 207724777-207724777 on Build GRCh38
rs987104965 Chr.2: 207724742-207724742 on Build GRCh38

View Full Product Details