Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127945320-127945569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM102A
Entrez Gene ID
399665
Gene Name
family with sequence similarity 102 member A
Gene Aliases
C9orf132, EEIG1, SYM-3A, bA203J24.7
UniGene
Hs.535972
Gene Chromosome Location
Chr.9: 127940582-127980533 on Build GRCh38
Transcripts
NM_001035254.2 NM_203305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1460165 c.859C>T p.P287S Chr.9: 127945425-127945425 on Build GRCh38
1205995 c.917+1G>A p.? Chr.9: 127945366-127945366 on Build GRCh38
1460166 c.848G>A p.R283Q Chr.9: 127945436-127945436 on Build GRCh38
1292795 c.835G>T p.E279* Chr.9: 127945449-127945449 on Build GRCh38
5530765 c.907C>T p.R303C Chr.9: 127945377-127945377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs554494022 Chr.9: 127945469-127945469 on Build GRCh38
rs748487907 Chr.9: 127945540-127945540 on Build GRCh38
rs752276424 Chr.9: 127945483-127945483 on Build GRCh38
rs571843903 Chr.9: 127945462-127945462 on Build GRCh38
rs766440485 Chr.9: 127945351-127945351 on Build GRCh38
rs770932996 Chr.9: 127945485-127945485 on Build GRCh38
rs529329211 Chr.9: 127945397-127945397 on Build GRCh38
rs1055421057 Chr.9: 127945475-127945475 on Build GRCh38
rs747931221 Chr.9: 127945378-127945378 on Build GRCh38
rs34532275 Chr.9: 127945408-127945408 on Build GRCh38
rs759485402 Chr.9: 127945365-127945365 on Build GRCh38
rs201753666 Chr.9: 127945381-127945381 on Build GRCh38
rs147692753 Chr.9: 127945512-127945512 on Build GRCh38
rs768795434 Chr.9: 127945536-127945536 on Build GRCh38
rs377579290 Chr.9: 127945421-127945421 on Build GRCh38
rs560284942 Chr.9: 127945396-127945396 on Build GRCh38
rs527578054 Chr.9: 127945377-127945377 on Build GRCh38
rs200989232 Chr.9: 127945436-127945436 on Build GRCh38
rs77325091 Chr.9: 127945514-127945514 on Build GRCh38
rs1001513189 Chr.9: 127945398-127945398 on Build GRCh38
rs776479474 Chr.9: 127945360-127945360 on Build GRCh38
rs746724491 Chr.9: 127945502-127945502 on Build GRCh38
rs762022952 Chr.9: 127945437-127945437 on Build GRCh38
rs780854071 Chr.9: 127945509-127945509 on Build GRCh38
rs777386502 Chr.9: 127945501-127945501 on Build GRCh38
rs370047060 Chr.9: 127945400-127945400 on Build GRCh38
rs567140266 Chr.9: 127945511-127945511 on Build GRCh38
rs750781284 Chr.9: 127945544-127945544 on Build GRCh38
rs770890330 Chr.9: 127945412-127945412 on Build GRCh38
rs201577073 Chr.9: 127945517-127945517 on Build GRCh38
rs762848563 Chr.9: 127945533-127945533 on Build GRCh38
rs749403138 Chr.9: 127945530-127945530 on Build GRCh38
rs2296705 Chr.9: 127945543-127945543 on Build GRCh38
rs760505505 Chr.9: 127945346-127945346 on Build GRCh38
rs944218024 Chr.9: 127945352-127945352 on Build GRCh38
rs771787971 Chr.9: 127945341-127945341 on Build GRCh38
rs531762368 Chr.9: 127945422-127945422 on Build GRCh38
rs369123963 Chr.9: 127945411-127945411 on Build GRCh38
rs953178126 Chr.9: 127945474-127945474 on Build GRCh38
rs370993152 Chr.9: 127945428-127945428 on Build GRCh38
rs867120716 Chr.9: 127945376-127945376 on Build GRCh38
rs767831626 Chr.9: 127945440-127945440 on Build GRCh38
rs373594027 Chr.9: 127945420-127945420 on Build GRCh38
rs771892560 Chr.9: 127945380-127945380 on Build GRCh38
rs762033195 Chr.9: 127945542-127945542 on Build GRCh38
rs569955360 Chr.9: 127945427-127945427 on Build GRCh38
rs750737789 Chr.9: 127945461-127945461 on Build GRCh38
rs866586915 Chr.9: 127945535-127945535 on Build GRCh38
rs962940079 Chr.9: 127945340-127945340 on Build GRCh38
rs574557228 Chr.9: 127945516-127945516 on Build GRCh38
rs376672692 Chr.9: 127945409-127945409 on Build GRCh38
rs764139297 Chr.9: 127945434-127945434 on Build GRCh38
rs751946480 Chr.9: 127945388-127945388 on Build GRCh38
rs774404788 Chr.9: 127945538-127945538 on Build GRCh38
rs565145117 Chr.9: 127945342-127945342 on Build GRCh38
rs200021618 Chr.9: 127945508-127945508 on Build GRCh38
rs775464690 Chr.9: 127945423-127945423 on Build GRCh38
rs111435518 Chr.9: 127945356-127945356 on Build GRCh38
rs746901159 Chr.9: 127945407-127945407 on Build GRCh38

View Full Product Details