Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19652518-19652791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP13A1
Entrez Gene ID
57130
Gene Name
ATPase 13A1
Gene Aliases
ATP13A, CGI-152
UniGene
Hs.501794
Gene Chromosome Location
Chr.19: 19645198-19663694 on Build GRCh38
Transcripts
NM_020410.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936722 c.1864T>C p.S622P Chr.19: 19652603-19652603 on Build GRCh38
6607242 c.2147T>C p.V716A Chr.19: 19652674-19652674 on Build GRCh38
4936721 c.2218T>C p.S740P Chr.19: 19652603-19652603 on Build GRCh38
7291344 c.1775A>G p.E592G Chr.19: 19652692-19652692 on Build GRCh38
3531139 c.2204A>G p.E735G Chr.19: 19652617-19652617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779026306 Chr.19: 19652578-19652578 on Build GRCh38
rs145358693 Chr.19: 19652609-19652609 on Build GRCh38
rs769132947 Chr.19: 19652769-19652769 on Build GRCh38
rs780916050 Chr.19: 19652716-19652716 on Build GRCh38
rs767748918 Chr.19: 19652628-19652628 on Build GRCh38
rs371018354 Chr.19: 19652579-19652579 on Build GRCh38
rs1033727553 Chr.19: 19652743-19652743 on Build GRCh38
rs200083832 Chr.19: 19652673-19652673 on Build GRCh38
rs544960226 Chr.19: 19652546-19652546 on Build GRCh38
rs901541588 Chr.19: 19652709-19652709 on Build GRCh38
rs369938079 Chr.19: 19652739-19652739 on Build GRCh38
rs533388585 Chr.19: 19652573-19652573 on Build GRCh38
rs774619676 Chr.19: 19652770-19652770 on Build GRCh38
rs375308930 Chr.19: 19652676-19652676 on Build GRCh38
rs775931797 Chr.19: 19652560-19652560 on Build GRCh38
rs373702947 Chr.19: 19652588-19652588 on Build GRCh38
rs769079462 Chr.19: 19652620-19652620 on Build GRCh38
rs763326424 Chr.19: 19652563-19652563 on Build GRCh38
rs758394802 Chr.19: 19652697-19652697 on Build GRCh38
rs764386744 Chr.19: 19652565-19652565 on Build GRCh38
rs758271260 Chr.19: 19652584-19652584 on Build GRCh38
rs942976937 Chr.19: 19652665-19652665 on Build GRCh38
rs760861293 Chr.19: 19652649-19652649 on Build GRCh38
rs750571841 Chr.19: 19652629-19652629 on Build GRCh38
rs564785652 Chr.19: 19652570-19652570 on Build GRCh38
rs918666397 Chr.19: 19652587-19652587 on Build GRCh38
rs374610173 Chr.19: 19652567-19652567 on Build GRCh38
rs763387344 Chr.19: 19652615-19652615 on Build GRCh38
rs754016963 Chr.19: 19652586-19652586 on Build GRCh38
rs966647504 Chr.19: 19652576-19652576 on Build GRCh38
rs1006729707 Chr.19: 19652726-19652726 on Build GRCh38
rs751337345 Chr.19: 19652712-19652712 on Build GRCh38
rs865930212 Chr.19: 19652602-19652602 on Build GRCh38
rs770251117 Chr.19: 19652555-19652555 on Build GRCh38
rs142504675 Chr.19: 19652704-19652704 on Build GRCh38
rs2288864 Chr.19: 19652717-19652717 on Build GRCh38
rs755006283 Chr.19: 19652574-19652574 on Build GRCh38
rs774768500 Chr.19: 19652621-19652621 on Build GRCh38
rs371636656 Chr.19: 19652556-19652556 on Build GRCh38
rs770595625 Chr.19: 19652596-19652596 on Build GRCh38
rs780602859 Chr.19: 19652597-19652597 on Build GRCh38
rs911363715 Chr.19: 19652612-19652612 on Build GRCh38
rs369144941 Chr.19: 19652548-19652548 on Build GRCh38
rs978360323 Chr.19: 19652577-19652577 on Build GRCh38
rs761707973 Chr.19: 19652647-19652648 on Build GRCh38
rs767555324 Chr.19: 19652571-19652571 on Build GRCh38
rs756958183 Chr.19: 19652714-19652714 on Build GRCh38
rs772366341 Chr.19: 19652653-19652654 on Build GRCh38
rs144490693 Chr.19: 19652733-19652734 on Build GRCh38
rs187111671 Chr.19: 19652627-19652627 on Build GRCh38
rs749855186 Chr.19: 19652734-19652734 on Build GRCh38
rs762865426 Chr.19: 19652741-19652742 on Build GRCh38
rs985199309 Chr.19: 19652585-19652585 on Build GRCh38
rs779537806 Chr.19: 19652724-19652724 on Build GRCh38
rs769241237 Chr.19: 19652604-19652604 on Build GRCh38

View Full Product Details